Learn More
Thermo Scientific Chemicals L-Arginine, 98+%
27.14 € - 560.79 €
Chemical Identifiers
CAS | 74-79-3 |
---|---|
Molecular Formula | C6H14N4O2 |
Molecular Weight (g/mol) | 174.20 |
MDL Number | MFCD00002635 |
InChI Key | ODKSFYDXXFIFQN-UHFFFAOYNA-N |
Synonym | l-arginine, arginine, l-+-arginine, l-arg, l +-arginine, s-2-amino-5-guanidinopentanoic acid, h-arg-oh, arginina, arginine, l |
PubChem CID | 6322 |
ChEBI | CHEBI:16467 |
SMILES | NC(CCCN=C(N)N)C(O)=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11498850
|
Thermo Scientific Chemicals
A15738.14 |
25 g |
27.14 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11402388
|
Thermo Scientific Chemicals
A15738.22 |
100 g |
52.99 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11412388
|
Thermo Scientific Chemicals
A15738.36 |
500 g |
156.78 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11422388
|
Thermo Scientific Chemicals
A15738.0E |
2500 g |
560.79 €
2500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
L-Arginine is used for heart and blood vessel conditions which includes congestive heart failure (CHF), chest pain, high blood pressure and coronary artery disease. It plays a vital role in the treatment of cardiovascular disease due to it being antiatherogenic, anti-ischemic, antiplatelet and antithrombotic. It acts as a growth stimulant and is involved in the treatment of erectile dysfunction in men. It is an important ingredient of tooth paste which provides effective relief for sensitive teeth.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
SolubilitySoluble in water. Slightly soluble in ethanol. Insoluble in ethyl ether
Notes
Air sensitive. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.
Chemical Identifiers
74-79-3 | |
174.20 | |
ODKSFYDXXFIFQN-UHFFFAOYNA-N | |
6322 | |
NC(CCCN=C(N)N)C(O)=O |
C6H14N4O2 | |
MFCD00002635 | |
l-arginine, arginine, l-+-arginine, l-arg, l +-arginine, s-2-amino-5-guanidinopentanoic acid, h-arg-oh, arginina, arginine, l | |
CHEBI:16467 |
Specifications
226°C (decomposition) | |
98+% | |
MFCD00002635 | |
14,780 | |
Soluble in water. Slightly soluble in ethanol. Insoluble in ethyl ether | |
NC(CCCN=C(N)N)C(O)=O | |
174.20 | |
CHEBI:16467 | |
+26° (c=1.6 in 6N HCl) | |
≥98% |
74-79-3 | |
C6H14N4O2 | |
1725413 | |
l-arginine, arginine, l-+-arginine, l-arg, l +-arginine, s-2-amino-5-guanidinopentanoic acid, h-arg-oh, arginina, arginine, l | |
ODKSFYDXXFIFQN-UHFFFAOYNA-N | |
2-amino-5-[(diaminomethylidene)amino]pentanoic acid | |
6322 | |
174.2 | |
Air Sensitive | |
L-Arginine |
Safety and Handling
P264b-P280i-P305+P351+P338
H319
missing translation for 'einecsNumber' : 200-811-1
missing translation for 'rtecsNumber' : CF1934200
missing translation for 'tsca' : Yes
missing translation for 'storageNote' : Air sensitive
Recommended Storage : Ambient temperatures; Store under Nitrogen
- L-Arginine
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
Hazard Statement
- H319-Causes serious eye irritation.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
- MIXTURE LIST-Contains : L-Arginine
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.