Learn More
Thermo Scientific™ L-Arginine-HCl, 13C6, 15N4 for SILAC
Study protein expression by mass spectrometry with mass variants of lysine, arginine and other amino acids to perform stable isotope labeling with amino acids in cell culture.
616.00 € - 3800.00 €
Specifications
For Use With (Equipment) | Mass Spectrometer |
---|---|
Product Type | Amino Acids |
Content And Storage | Store at room temperature. |
Label or Dye | Mass Isotope of Amino Acid |
Shipping Condition | Room Temperature |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10065703
|
Thermo Scientific™
89990 |
50 mg |
616.00 €
50mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
12166480
|
Thermo Scientific™
88434 |
500 mg |
3800.00 €
500mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Beschreibung
Thermo Scientific™ Heavy and Light Amino Acids are used to specifically analyze protein expression by mass spectrometry using stable isotope labeling with amino acids in cell culture (SILAC) quantification kits.
SILAC is a powerful method to identify and quantify relative differential changes in complex protein samples. This approach entails the in vivo metabolic incorporation of “heavy“ 13C- or 15N-labeled amino acids into proteins followed by mass spectrometry (MS) for the accelerated and comprehensive identification, characterization and quantitation of proteins.Highlights:
Efficient – 100% label incorporation into proteins of living cells
Flexible – different isotopes of heavy and/or light amino acids for arginine, lysine, leucine and proline enable the quantitation of peptides derived from MS-grade proteases
Multiplex capabilities – several alternative isotopes of arginine and lysine are available that allow the analysis of multiple treatment conditions in each experiment
High-quality supplements – heavy amino acids with >99% isotope purity
Spezifikation
Mass Spectrometer | |
Store at room temperature. | |
Room Temperature | |
Protein | |
Protein Labeling |
Amino Acids | |
Mass Isotope of Amino Acid | |
Cells | |
Mass Spectrometry | |
Solid |
Sicherheit und Handhabung
- L-Arginine-HCl, 13C6, 15N4 for SILAC
Signalwort
- Warning
Gefahrenkategorie
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
Gefahrenhinweis
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
Sicherheitshinweis
- P264-Wash hands thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
- P337+P313-If eye irritation persists: Get medical advice/attention.
- P362+P364-Take off contaminated clothing and wash it before reuse.
Ergänzende Informationen
- MIXTURE LIST-Contains : (+)-L-arginine hydrochloride
For Research Use Only. Not for use in diagnostic procedures.
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.