missing translation for 'onlineSavingsMsg'
Learn More

L-Arginine Hydrochloride (White Crystals or Crystalline Powder), Fisher BioReagents

Product Code. 10061503
Click to view available options
Quantity:
100 g
Unit Size:
100g
This item is not returnable. View return policy

Product Code. 10061503

Brand: Fisher BioReagents BP372100

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
The hydrochloride salt of L-Arginine is suitable for use in tissue culture systems requiring additives and has many of the same applications as L-Arginine.
TRUSTED_SUSTAINABILITY

Specifications

Melting Point 226°C
Ammonium 0.02% max.
CAS 1119-34-2
Boiling Point 235°C
Color White
Chloride 16.58 to 17.00%
Heavy Metals (as Pb) 10ppm max.
Ignition Residue 0.1% max.
Identification Pass Test
Assay Percent Range 98.5 to 101.0 %
Loss on Drying 0.2% max.
Other Amino Acids Not Detected
Molecular Formula C6H15ClN4O2
Packaging Glass Bottle
Specific Rotation 22 to 23° (+ or -)
Merck Index 15, 770
State of Solution (%T) 98.0% min.
Synonym l-arginine hydrochloride, arginine hydrochloride, l-arginine hcl, l-arginine monohydrochloride, h-arg-oh.hcl, r-gene, arginine monohydrochloride, argamine, l-arginine, monohydrochloride, unii-f7lth1e20y
Alpha Vector Arginine Hydrochloride
InChI Key KWTQSFXGGICVPE-WCCKRBBISA-N
SMILES C(CC(C(=O)O)N)CN=C(N)N.Cl
IUPAC Name (2S)-2-amino-5-(diaminomethylideneamino)pentanoic acid;hydrochloride
Molecular Weight (g/mol) 210.662
Quantity 100 g
PubChem CID 66250
Formula Weight 210.58
Percent Purity 98.5 to 101.0%
Physical Form Solid
Chemical Name or Material L-Arginine Hydrochloride
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.