missing translation for 'onlineSavingsMsg'
Learn More

L-Arginine, Multi-Compendial, U.S.P., J.T. Baker™

Product Code. 15527954
Click to view available options
Quantity:
1 kg
Packaging:
Poly Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Specifications

CAS 74-79-3
Molecular Formula C6H14N4O2
MDL Number MFCD00002635
Packaging Poly Bottle
Synonym l-arginine, arginine, l-+-arginine, l-arg, l +-arginine, s-2-amino-5-guanidinopentanoic acid, h-arg-oh, arginina, arginine, l
InChI Key ODKSFYDXXFIFQN-UHFFFAOYNA-N
SMILES NC(CCCN=C(N)N)C(O)=O
IUPAC Name 2-amino-5-[(diaminomethylidene)amino]pentanoic acid
Molecular Weight (g/mol) 174.20
PubChem CID 6322
ChEBI CHEBI:16467
Formula Weight 174.2
Grade Multi-Compendial/USP
Quantity 1 kg
Chemical Name or Material L-Arginine
Show More Show Less
compliance-icons
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : L-Arginine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'