missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals L-Ascorbic acid sodium salt, 99%

Product Code. 10012011 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
100 g
500 g
2.5 kg
Packaging:
Glass bottle
Plastic bottle
Unit Size:
100g
2.5kg
500g
5g
This item is not returnable. View return policy

Product Code. 10012011

Brand: Thermo Scientific Chemicals undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 134-03-2 | C6H7NaO6 | 198.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 134-03-2
Molecular Formula C6H7NaO6
Molecular Weight (g/mol) 198.11
MDL Number MFCD00082340
InChI Key IFVCRSPJFHGFCG-HXPAKLQESA-N
Synonym sodium ascorbate, l-ascorbic acid sodium salt, sodium l-ascorbate, vitamin c sodium, ascorbicin, sodascorbate, cebitate, aminofenitrooxon, natrii ascorbas, monosodium l-ascorbate
PubChem CID 131674100
IUPAC Name sodium 5-[(1S)-1,2-dihydroxyethyl]-3-hydroxy-2,4-dioxooxolan-3-ide
SMILES [Na+].OC[C@H](O)C1OC(=O)[C-](O)C1=O

Specifications

Melting Point 219.0°C to 221.0°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 98.5 to 101.5% (Iodimetry)
Packaging Glass bottle
Quantity 5 g
Merck Index 15,819
Specific Rotation + 104.00
Solubility Information (5% in water) Clear colorless to light yellow
Formula Weight 198.11
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material L-Ascorbic acid sodium salt, 99%
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.