missing translation for 'onlineSavingsMsg'
Learn More

L-Ascorbic acid, Tocris Bioscience™

Product Code. 18063932
Click to view available options
Quantity:
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Enhances the generation of iPSCs; increases reprogramming efficiency

Inhibitor of Cav3.2 channels (IC50 = 6.5 μM); displays no effect on Cav3.1 or Cav3.3 channels heterologously expressed in HEK 293 cells. Also enhances the generation of induced pluripotent stem cells (iPSCs) from mouse and human somatic cells by increasing reprogramming efficiency. Naturally occurring antioxidant.

Specifications

CAS 50-81-7
Molecular Formula C6H8O6
MDL Number MFCD00064328
Quantity 50 mg
Synonym l-ascorbic acid, ascorbic acid, vitamin c, l-ascorbate, ascorbate, ascorbicap, l +-ascorbic acid, cevitamic acid, ascoltin, hybrin
InChI Key CIWBSHSKHKDKBQ-DOMZIZNONA-N
SMILES OC[C@H](O)[C@H]1OC(=O)C(O)=C1O
IUPAC Name (5R)-5-[(1S)-1,2-dihydroxyethyl]-3,4-dihydroxy-2,5-dihydrofuran-2-one
Molecular Weight (g/mol) 176.12
PubChem CID 54670067
ChEBI CHEBI:29073
Formula Weight 176.12
Percent Purity >99%
Chemical Name or Material L-Ascorbic acid
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'