missing translation for 'onlineSavingsMsg'
Learn More
Thermo Scientific Chemicals L(+)-Asparagine monohydrate, 99% Product Code.: 10164430

Thermo Scientific Chemicals L(+)-Asparagine monohydrate, 99%

Product Code. 10164430
100 g, Glass Bottle
Click to view available options
Quantity:
100 g
250 g
1 kg
5 kg
Packaging:
Glass Bottle
Plastic Bottle
This item is not returnable. View return policy

Product Code. 10164430

Brand: Thermo Scientific Chemicals 175271000

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

L(+)-Asparagine monohydrate, 99%, C4H10N2O4, CAS Number-5794-13-8, asparagine monohydrate, l +-asparagine monohydrate, asparagine, asparagine nf, l-asparagine hydrate, h-asn-oh.h2o, unii-2pd79vf521, l-asparagine monohydrate, l-asparagine, monohydrate, s-2,4-diamino-4-oxobutanoic acid hydrate, 250g

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5794-13-8
Molecular Formula C4H10N2O4
Molecular Weight (g/mol) 150.134
MDL Number MFCD00151038
InChI Key RBMGJIZCEWRQES-DKWTVANSSA-N
Synonym l-asparagine monohydrate, l +-asparagine monohydrate, l-asparagine hydrate, asparagine monohydrate, h-asn-oh.h2o, l-asparagine, monohydrate, asparagine nf, s-2,4-diamino-4-oxobutanoic acid hydrate, asparagine, unii-2pd79vf521
PubChem CID 170358
IUPAC Name (2S)-2,4-diamino-4-oxobutanoic acid;hydrate
SMILES C(C(C(=O)O)N)C(=O)N.O

Specifications

Melting Point 233.0°C to 235.0°C
Ammonium 1000ppm max.
Arsenic (As) 1ppm max.
Color White
Chlorine (Cl) 210ppm max.
Heavy Metals (as Pb) 10ppm max.
Infrared Spectrum Authentic
Iron (Fe) 10ppm max.
Assay Percent Range 99%
Loss on Drying 11.5% to 12.5% (130°C, 3 hrs)
Other Amino Acids 1% max. (TLC)
Packaging Glass Bottle
Linear Formula H2NCOCH2CH(NH2)CO2H·H2O
Specific Rotation + 35.00
Sulfate 280ppm max.
Transmittance 98% min. (c=2, H2O, 430nm) 1cm cell
Sulfated Ash 0.1% max.
Solubility Information Solubility in water: 30g/L water (20°C). Other solubilities: soluble in acids and alkalies, practically insoluble in methanol, ethanol, ether, and benzene
Quantity 100 g
Formula Weight 150.14
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material L(+)-Asparagine monohydrate, 99%
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0443183 Certificate TypeCertificate of Analysis Date27/12/2023 Catalog Number175272500,175270010,175271000,175270050
Lot NumberA0432751 Certificate TypeCertificate of Analysis Date08/02/2023 Catalog Number175272500,175270010,175271000,175270050
Lot NumberA0402835 Certificate TypeCertificate of Analysis Date15/12/2022 Catalog Number175272500,175270010,175271000,175270050
Lot NumberA0403269 Certificate TypeCertificate of Analysis Date15/12/2022 Catalog Number175272500,175270010,175271000,175270050
Lot NumberA0398744 Certificate TypeCertificate of Analysis Date14/12/2022 Catalog Number175272500,175270010,175271000,175270050
Product Identifier
  • L(+)-Asparagine monohydrate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific Chemicals L(+)-Asparagine monohydrate, 99% >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.