missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L(+)-Asparagine monohydrate, Thermo Scientific Chemicals
53.77 € - 414.06 €
Chemical Identifiers
CAS | 5794-13-8 |
---|---|
Molecular Formula | C4H10N2O4 |
Molecular Weight (g/mol) | 150.134 |
InChI Key | RBMGJIZCEWRQES-DKWTVANSSA-N |
Synonym | l-asparagine monohydrate, l +-asparagine monohydrate, l-asparagine hydrate, asparagine monohydrate, h-asn-oh.h2o, l-asparagine, monohydrate, asparagine nf, s-2,4-diamino-4-oxobutanoic acid hydrate, asparagine, unii-2pd79vf521 |
PubChem CID | 170358 |
IUPAC Name | (2S)-2,4-diamino-4-oxobutanoic acid;hydrate |
SMILES | C(C(C(=O)O)N)C(=O)N.O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15259295
|
Thermo Scientific Acros
401481000 |
100 g |
53.77 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15269295
|
Thermo Scientific Acros
401482500 |
250 g |
112.56 €
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15249295
|
Thermo Scientific Acros
401480010 |
1 kg |
414.06 €
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
5794-13-8 | |
150.134 | |
l-asparagine monohydrate, l +-asparagine monohydrate, l-asparagine hydrate, asparagine monohydrate, h-asn-oh.h2o, l-asparagine, monohydrate, asparagine nf, s-2,4-diamino-4-oxobutanoic acid hydrate, asparagine, unii-2pd79vf521 | |
(2S)-2,4-diamino-4-oxobutanoic acid;hydrate |
C4H10N2O4 | |
RBMGJIZCEWRQES-DKWTVANSSA-N | |
170358 | |
C(C(C(=O)O)N)C(=O)N.O |
Specifications
233.0°C to 235.0°C | |
C4H10N2O4 | |
l-asparagine monohydrate, l +-asparagine monohydrate, l-asparagine hydrate, asparagine monohydrate, h-asn-oh.h2o, l-asparagine, monohydrate, asparagine nf, s-2,4-diamino-4-oxobutanoic acid hydrate, asparagine, unii-2pd79vf521 | |
RBMGJIZCEWRQES-DKWTVANSSA-N | |
(2S)-2,4-diamino-4-oxobutanoic acid;hydrate | |
150.134 | |
150.14 |
5794-13-8 | |
H2NCOCH2CH(NH2)CO2H·H2O | |
Solubility in water: 30g/L water (20°C). Other solubilities: soluble in acids and alkalies, practically insoluble in methanol, ethanol, ether, and benzene | |
C(C(C(=O)O)N)C(=O)N.O | |
+ 35.00 | |
170358 | |
L(+)-Asparagine monohydrate |
Safety and Handling
Product Identifier
- L(+)-Asparagine monohydrate
RUO – Research Use Only