missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals L-Aspartic acid, 98+%
30.20 € - 345.00 €
Chemical Identifiers
CAS | 56-84-8 |
---|---|
Molecular Formula | C4H7NO4 |
Molecular Weight (g/mol) | 133.10 |
MDL Number | MFCD00002616 |
InChI Key | CKLJMWTZIZZHCS-UHFFFAOYNA-N |
Synonym | l-aspartic acid, aspartic acid, h-asp-oh, l-aspartate, asparagic acid, aspatofort, l-asparagic acid, l-aminosuccinic acid, s-2-aminosuccinic acid, asparaginic acid |
PubChem CID | 5960 |
ChEBI | CHEBI:17053 |
SMILES | NC(CC(O)=O)C(O)=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11445623
|
Thermo Scientific Chemicals
A13520.30 |
250 g |
30.20 €
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11435623
|
Thermo Scientific Chemicals
A13520.0B |
1000 g |
91.70 €
1000g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11455623
|
Thermo Scientific Chemicals
A13520.0I |
5000 g |
345.00 €
5000g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
L-Aspartic acid is used in enzyme active centers, as well as in maintaining the solubility and ionic character of proteins.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsL-Aspartic acid is used in enzyme active centers, as well as in maintaining the solubility and ionic character of proteins.
Solubility
Soluble in water(4.5g/L).
Notes
Store in cool, dry conditions in well sealed containers. Keep container tightly closed.
Chemical Identifiers
56-84-8 | |
133.10 | |
CKLJMWTZIZZHCS-UHFFFAOYNA-N | |
5960 | |
NC(CC(O)=O)C(O)=O |
C4H7NO4 | |
MFCD00002616 | |
l-aspartic acid, aspartic acid, h-asp-oh, l-aspartate, asparagic acid, aspatofort, l-asparagic acid, l-aminosuccinic acid, s-2-aminosuccinic acid, asparaginic acid | |
CHEBI:17053 |
Specifications
>300°C (decomposition) | |
C4H7NO4 | |
1723530 | |
l-aspartic acid, aspartic acid, h-asp-oh, l-aspartate, asparagic acid, aspatofort, l-asparagic acid, l-aminosuccinic acid, s-2-aminosuccinic acid, asparaginic acid | |
CKLJMWTZIZZHCS-UHFFFAOYNA-N | |
(2S)-2-aminobutanedioic acid | |
5960 | |
133.1 | |
1.66 | |
+25° (c=2 in 5N HCl) |
56-84-8 | |
MFCD00002616 | |
14,840 | |
Soluble in water(4.5g/L). | |
NC(CC(O)=O)C(O)=O | |
133.10 | |
CHEBI:17053 | |
≥98% | |
Odorless | |
L-Aspartic acid |
Sécurité et traitement
missing translation for 'einecsNumber' : 200-291-6
missing translation for 'rtecsNumber' : CI9098500
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only