missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals L-Aspartyl-L-phenylalanine, 95%

Product Code. 10647061
100 mg, Glass Bottle
Click to view available options
Quantity:
100 mg
500 mg
Packaging:
Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13433-09-5
Molecular Formula C13H16N2O5
Molecular Weight (g/mol) 280.28
MDL Number MFCD00063155
InChI Key YZQCXOFQZKCETR-UWVGGRQHSA-N
Synonym l-aspartyl-l-phenylalanine, aspartyl-phenylalanine, demethylaspartame, aspartylphenylalanine, asp-phe, h-asp-phe-oh, l-a-aspartyl-l-phenylalanine, l-alpha-aspartyl-l-phenylalanine, alpha-aspartylphenylalanine, s-3-amino-4-s-1-carboxy-2-phenylethyl amino-4-oxobutanoic acid
PubChem CID 93078
ChEBI CHEBI:73830
SMILES N[C@@H](CC(O)=O)C(=O)N[C@@H](CC1=CC=CC=C1)C(O)=O

Specifications

Color Tan to White
Infrared Spectrum Authentic
Assay Percent Range 95%
Packaging Glass Bottle
Linear Formula HO2CCH2CH(NH2)CONHCH(CH2C6H5)CO2H
Specific Rotation +10.0° to +13.0° (24°C, 589nm) (c=1, 0.5N HCl)
Water 3% max. (K.F.)
IUPAC Name (3S)-3-amino-3-{[(1S)-1-carboxy-2-phenylethyl]carbamoyl}propanoic acid
Quantity 100 mg
Formula Weight 280.28
Percent Purity ≥94% (HPLC)
Physical Form Powder
Chemical Name or Material L-Aspartyl-L-phenylalanine, 95%
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific Chemicals L-Aspartyl-L-phenylalanine, 95% >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.