missing translation for 'onlineSavingsMsg'
Learn More

L-Cysteine hydrochloride, MedChemExpress

MedChemExpress L-Cysteine hydrochloride is a conditionally essential amino acid, which acts as a precursor for biologically active molecules such as hydrogen sulphide (H2S), glutathione and taurine. L-Cysteine hydrochloride suppresses ghrelin and reduces appetite in rodents and humans.

Brand:  Medchem Express HY-Y0337A

Encompass_Preferred

Product Code. 16433469

  • 79.65 € / 1g

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

L-Cysteine hydrochloride is a conditionally essential amino acid, which acts as a precursor for biologically active molecules such as hydrogen sulphide (H2S), glutathione and taurine. L-Cysteine hydrochloride suppresses ghrelin and reduces appetite in rodents and humans.
Specifications

Specifications

L-Cysteine hydrochloride
White to Off-White
C3H8ClNO2S
H2O : 100 mg/mL (634.44 mM; Need ultrasonic)
157.62
97.0%
Solid
52-89-1
Research
1 g
N[C@@H](CS)C(O)=O.[H]Cl
157.62
Research
Metabolism-protein/nucleotide metabolism
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Research purposes only