missing translation for 'onlineSavingsMsg'
Learn More

L-Cysteine, MedChemExpress

MedChemExpress L-Cysteine is a conditionally essential amino acid, which acts as a precursor for biologically active molecules such as hydrogen sulphide (H2S), glutathione and taurine. L-Cysteine suppresses ghrelin and reduces appetite in rodents and humans.

Brand:  Medchem Express HY-Y0337

Encompass_Preferred

Product Code. 16423469

  • 79.65 € / 1g
Estimated Shipment: 03-12-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

L-Cysteine is a conditionally essential amino acid, which acts as a precursor for biologically active molecules such as hydrogen sulphide (H2S), glutathione and taurine. L-Cysteine suppresses ghrelin and reduces appetite in rodents and humans.
Specifications

Specifications

L-Cysteine
White
C3H7NO2S
H2O : 33.33 mg/mL (275.09 mM; Need ultrasonic)
121.16
95.0%
Solid
52-90-4
Research
1 g
N[C@@H](CS)C(O)=O
121.16
Research
Metabolism-protein/nucleotide metabolism
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Research purposes only