missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals L-Cysteine methyl ester hydrochloride
Click to view available options
Quantity:
25 g
100 g
250 g
Description
L-Cysteine methyl ester hydrochloride is a biochemical shown to inhibit binding of ethynylestradiol metabolites to protein and nucleic acids. It used in the synthesis of peptides, Preparation of thiazolidine-4-carboxylates with carbonyl compounds.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
L-Cysteine methyl ester hydrochloride is a biochemical shown to inhibit binding of ethynylestradiol metabolites to protein and nucleic acids. It used in the synthesis of peptides, Preparation of thiazolidine-4-carboxylates with carbonyl compounds.
Solubility
Soluble in water
Notes
Protect from heat. Store away from oxidizing agents.
L-Cysteine methyl ester hydrochloride is a biochemical shown to inhibit binding of ethynylestradiol metabolites to protein and nucleic acids. It used in the synthesis of peptides, Preparation of thiazolidine-4-carboxylates with carbonyl compounds.
Solubility
Soluble in water
Notes
Protect from heat. Store away from oxidizing agents.
Specifications
Specifications
Melting Point | 142°C (decomposition) |
Color | White |
Beilstein | 3685824 |
Merck Index | 14,5785 |
Solubility Information | Soluble in water |
Formula Weight | 171.65 |
Quantity | 25 g |
Physical Form | Crystalline Powder |
Chemical Name or Material | L-Cysteine methyl ester hydrochloride |
Safety and Handling
Signal Word
- Warning
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'