missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals L-Cysteine methyl ester hydrochloride

Product Code. 15496379
Click to view available options
Quantity:
25 g
100 g
250 g
This item is not returnable. View return policy
This item is not returnable. View return policy

L-Cysteine methyl ester hydrochloride is a biochemical shown to inhibit binding of ethynylestradiol metabolites to protein and nucleic acids. It used in the synthesis of peptides, Preparation of thiazolidine-4-carboxylates with carbonyl compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
L-Cysteine methyl ester hydrochloride is a biochemical shown to inhibit binding of ethynylestradiol metabolites to protein and nucleic acids. It used in the synthesis of peptides, Preparation of thiazolidine-4-carboxylates with carbonyl compounds.

Solubility
Soluble in water

Notes
Protect from heat. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 18598-63-5
Molecular Formula C4H10ClNO2S
Molecular Weight (g/mol) 171.639
MDL Number MFCD00038998
InChI Key WHOHXJZQBJXAKL-DFWYDOINSA-N
Synonym l-cysteine methyl ester hydrochloride, methyl l-cysteinate hydrochloride, h-cys-ome.hcl, l-cystein methyl ester hydrochloride, acdrile, actiol, mecysteine hydrochloride, cysteine methyl ester hydrochloride, r-methyl 2-amino-3-mercaptopropanoate hydrochloride
PubChem CID 2733208
IUPAC Name methyl (2R)-2-amino-3-sulfanylpropanoate;hydrochloride
SMILES COC(=O)C(CS)N.Cl

Specifications

Melting Point 142°C (decomposition)
Color White
Beilstein 3685824
Merck Index 14,5785
Solubility Information Soluble in water
Quantity 100 g
Formula Weight 171.65
Physical Form Crystalline Powder
Chemical Name or Material L-Cysteine methyl ester hydrochloride

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.