missing translation for 'onlineSavingsMsg'
Learn More

L-Cystine, Multi-Compendial, F.C.C., J.T. Baker™

Product Code. 15577954
Click to view available options
Quantity:
1 kg
Packaging:
Poly Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Specifications

CAS 56-89-3
Molecular Formula C6H12N2O4S2
Packaging Poly Bottle
Synonym l-cystine, cystine, l-dicysteine, l-cystin, cystine acid, cysteine disulfide, oxidized l-cysteine, beta,beta'-dithiodialanine, h-cys-oh 2, l-cysteine disulfide
InChI Key LEVWYRKDKASIDU-IMJSIDKUSA-N
SMILES C(C(C(=O)O)N)SSCC(C(=O)O)N
IUPAC Name (2R)-2-amino-3-[[(2R)-2-amino-2-carboxyethyl]disulfanyl]propanoic acid
Molecular Weight (g/mol) 240.292
Quantity 1 kg
PubChem CID 67678
ChEBI CHEBI:16283
Formula Weight 240.3
Grade FCC/Multi-Compendial
Chemical Name or Material L-Cystine
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.