missing translation for 'onlineSavingsMsg'
Learn More

L-DOPA, Tocris Bioscience™

Product Code. 18722494
Click to view available options
Quantity:
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Dopamine precursor

Immediate precursor of dopamine, produced by tyrosine hydroxylase. Displays antiParkinsonian activity.

Specifications

CAS 59-92-7
Molecular Formula C9H11NO4
MDL Number MFCD00002598
Quantity 50 mg
Synonym levodopa, l-dopa, dopar, 3-hydroxy-l-tyrosine, 3,4-dihydroxy-l-phenylalanine, larodopa, bendopa, levopa, dihydroxy-l-phenylalanine, cidandopa
InChI Key WTDRDQBEARUVNC-LURJTMIESA-N
SMILES N[C@@H](CC1=CC=C(O)C(O)=C1)C(O)=O
IUPAC Name (2S)-2-amino-3-(3,4-dihydroxyphenyl)propanoic acid
Molecular Weight (g/mol) 197.19
PubChem CID 6047
ChEBI CHEBI:15765
Formula Weight 197.19
Percent Purity >99%
Chemical Name or Material L-DOPA
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'