Learn More
L(-)-Epinephrine, 99%, Thermo Scientific Chemicals
36.18 € - 415.07 €
Chemical Identifiers
CAS | 51-43-4 |
---|---|
MDL Number | MFCD00002204 |
InChI Key | UCTWMZQNUQWSLP-VIFPVBQESA-N |
Synonym | epinephrine, l-adrenaline, adrenaline, l-epinephrine, adrenalin, levoepinephrine, epinefrina, --adrenaline, adnephrine, chelafrin |
PubChem CID | 5816 |
ChEBI | CHEBI:28918 |
IUPAC Name | 4-[(1R)-1-hydroxy-2-(methylamino)ethyl]benzene-1,2-diol |
SMILES | CNCC(C1=CC(=C(C=C1)O)O)O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10219883
|
Thermo Scientific Acros
204400010 |
1 g | Glass bottle |
36.18 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10434253
|
Thermo Scientific Acros
204400050 |
5 g | Glass bottle |
114.57 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10740965
|
Thermo Scientific Acros
204400250 |
25 g | Glass bottle |
415.07 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
51-43-4 | |
UCTWMZQNUQWSLP-VIFPVBQESA-N | |
5816 | |
4-[(1R)-1-hydroxy-2-(methylamino)ethyl]benzene-1,2-diol |
MFCD00002204 | |
epinephrine, l-adrenaline, adrenaline, l-epinephrine, adrenalin, levoepinephrine, epinefrina, --adrenaline, adnephrine, chelafrin | |
CHEBI:28918 | |
CNCC(C1=CC(=C(C=C1)O)O)O |
Specifications
51-43-4 | |
White to Beige | |
Authentic | |
C9H13NO3 | |
MFCD00002204 | |
15, 3674 | |
− 51.50 | |
Solubility in water: sparingly soluble. Other solubilities: insoluble in alcohol, chloroform, ether and, acetone | |
CNCC(C1=CC(=C(C=C1)O)O)O | |
183.21 | |
CHEBI:28918 | |
99% | |
L(-)-Epinephrine, 99% |
208.0°C to 211.0°C | |
1% max. (18 hrs) (0.7 kPa, P2O5) | |
99% | |
(HO)2C6H3CH(CH2NHCH3)OH | |
13, 830 | |
− 51.50 (20.00°C c=4,1M HCl,dry sub) | |
epinephrine, l-adrenaline, adrenaline, l-epinephrine, adrenalin, levoepinephrine, epinefrina, --adrenaline, adnephrine, chelafrin | |
UCTWMZQNUQWSLP-VIFPVBQESA-N | |
4-[(1R)-1-hydroxy-2-(methylamino)ethyl]benzene-1,2-diol | |
5816 | |
183.21 | |
Fine Crystalline Powder |
Safety and Handling
GHS H Statement
Causes skin irritation. Toxic if inhaled. Toxic if swallowed. Toxic in contact with skin. Causes serious eye irritation. May cause respiratory irritation. Harmful to aquatic life with long lasting effects. May cause damage to organs through prolonged or repeated exposure.
GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician. Wear protective gloves/protective clothing/eye protection/face protection. Call a POISON CENTER or doctor/physician if you feel unwell. IF ON SKIN: Gently wash with plenty of soap and water. IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing. Avoid breathing dust/fume/gas/mist/vapors/spray. Do not breathe dust/fume/gas/mist/vapors/spray. Avoid release to the environment. IF ON SKIN: Wash with plenty of soap and water. Wear protective gloves/protective clothing/eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 200-098-7
missing translation for 'rtecsNumber' : DO2625000
missing translation for 'tsca' : TSCA
Recommended Storage : product darkens on exposure to air and light
- L(-)-Epinephrine
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 3
- LONG-TERM AQUATIC HAZARD Chronic 3
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity after repeated exposure Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H301-Toxic if swallowed.
- H311-Toxic in contact with skin.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H331-Toxic if inhaled.
- H335-May cause respiratory irritation.
- H373-May cause damage to organs.
- H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
- MIXTURE LIST-Contains : Epinephrine
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.