missing translation for 'onlineSavingsMsg'
Learn More
Thermo Scientific Chemicals L(+)-Glutamic acid, 99% Product Code.: 10458880

Thermo Scientific Chemicals L(+)-Glutamic acid, 99%

Product Code. 10458880
1 kg, Plastic Bottle
Click to view available options
Quantity:
100 g
250 g
1 kg
5 kg
Packaging:
Plastic Bottle
Plastic Drum
This item is not returnable. View return policy

Product Code. 10458880

Brand: Thermo Scientific Chemicals 156210010

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 56-86-0
Molecular Formula C5H9NO4
Molecular Weight (g/mol) 147.13
MDL Number MFCD00002634
InChI Key WHUUTDBJXJRKMK-UHFFFAOYNA-N
Synonym l-glutamic acid, glutamic acid, 2s-2-aminopentanedioic acid, h-glu-oh, s-2-aminopentanedioic acid, glutaminol, l-glutaminic acid, glutacid, glutamicol, glutamidex
PubChem CID 33032
ChEBI CHEBI:16015
IUPAC Name (2S)-2-aminopentanedioic acid
SMILES NC(CCC(O)=O)C(O)=O

Specifications

Ammonium 200ppm max.
Color White
Chlorine (Cl) 200ppm max.
Heavy Metals (as Pb) 10ppm max.
Infrared Spectrum Authentic
Iron (Fe) 30ppm max.
Assay Percent Range 99%
Loss on Drying 0.5% (105°C, 3 hrs) max.
Other Amino Acids Not detectable
Packaging Plastic Bottle
Linear Formula HO2CCH2CH2CH(NH2)CO2H
Specific Rotation +30.5° to +32.5° (20°C, 589nm) (on dried substance) (c=10, 2N HCl)
Sulfate 300ppm max.
Sulfated Ash 0.1% max.
Sublimation Point >200
Specific Rotation Condition +32.00 (20.00°C c=10,2N HCl)
Quantity 1 kg
Formula Weight 147.13
Percent Purity 98.5 to 100.5% (on dried substance)
Physical Form Powder
Chemical Name or Material L(+)-Glutamic acid
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific Chemicals L(+)-Glutamic acid, 99% >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.