missing translation for 'onlineSavingsMsg'
Learn More

L-Glutamic acid diethyl ester hydrochloride, 98%, Thermo Scientific Chemicals

Product Code. 15455908
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1118-89-4
Molecular Formula C9H18ClNO4
Molecular Weight (g/mol) 239.696
MDL Number MFCD00012509
InChI Key WSEQLMQNPBNMSL-FJXQXJEOSA-N
Synonym l-glutamic acid diethyl ester hydrochloride, diethyl l-glutamate hydrochloride, h-glu oet-oet.hcl, diethyl glutamate hydrochloride, h-glu oet-oet hcl, s-diethyl 2-aminopentanedioate hydrochloride, diethyl l-glutaminate hydrochloride, 1,5-diethyl 2s-2-aminopentanedioate hydrochloride, l-glutamic acid, diethyl ester, hydrochloride, diethyl 2s-2-aminopentanedioate hydrochloride
PubChem CID 73960
IUPAC Name diethyl (2S)-2-aminopentanedioate;hydrochloride
SMILES CCOC(=O)CCC(C(=O)OCC)N.Cl

Specifications

Melting Point 108°C to 110°C
Linear Formula C2H5OCOCH2CH2CH(NH2)COOC2H5·HCl
Beilstein 214-270-
Quantity 5 g
Formula Weight 239.7
Optical Rotation +22° (c=1 in H2O)
Percent Purity 98%
Chemical Name or Material L-Glutamic acid diethyl ester hydrochloride

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'