missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva L-Glutamine
Supplement serum- and protein-free media using this amino acid.
32.55 € - 37.20 €
Specifications
Content And Storage | Store frozen |
---|---|
Concentration | 29.2 mg/mL |
Form | Liquid |
Sterilization Method | 0.1 μm Sterile-filtered |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10453332
|
Cytiva
SH30034.01 |
100 mL |
32.55 €
100mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10691233
|
Cytiva
SH30034.02 |
500 mL |
37.20 €
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- Format: Liquid
Specifications
Store frozen | |
29.2 mg/mL | |
Liquid | |
0.1 μm Sterile-filtered |
Safety and Handling
missing translation for 'shelfLife' : 12 Months
Hazard Statement
- EUH210-Safety data sheet available on request.
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title