Learn More
L-Glutamine, 99%, Thermo Scientific Chemicals
Involved in many metabolic processes and regulates acid-base balance in the kidney. It is an important amino acid for the incorporation of ammonium ions into biomolecules. It is a vital component of culture media that serves as a major energy source for cells in culture.
40.00 € - 336.68 €
Chemical Identifiers
CAS | 56-85-9 |
---|---|
Molecular Formula | C5H10N2O3 |
Molecular Weight (g/mol) | 146.15 |
MDL Number | MFCD00008044 |
InChI Key | ZDXPYRJPNDTMRX-VKHMYHEASA-N |
Synonym | l-glutamine, glutamine, levoglutamide, l-+-glutamine, glutamic acid amide, h-gln-oh, stimulina, cebrogen, glumin, levoglutamid |
PubChem CID | 5961 |
ChEBI | CHEBI:18050 |
SMILES | N[C@@H](CCC(N)=O)C(O)=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11482277
|
Thermo Scientific Alfa Aesar
A14201.18 |
50 g |
40.00 €
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11492277
|
Thermo Scientific Alfa Aesar
A14201.30 |
250 g |
109.55 €
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11472277
|
Thermo Scientific Alfa Aesar
A14201.0B |
1000 g |
336.68 €
1000g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
L-Glutamine is involved in many metabolic processes and regulates acid-base balance in the kidney. It is an important amino acid for the incorporation of ammonium ions into biomolecules. It is a vital component of culture media that serves as a major energy source for cells in culture. As a component in oral supplementation, it is useful for patients who have advanced stages of cancer to reverse cachexia (i.e. muscle wasting). It is also used for attention deficit-hyperactivity disorder (ADHD), a urinary condition called cystinuria, sickle cell anemia and for alcohol withdrawal support. It finds applications in patients with HIV (AIDS) to prevent weight loss.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsL-Glutamine is involved in many metabolic processes and regulates acid-base balance in the kidney. It is an important amino acid for the incorporation of ammonium ions into biomolecules. It is a vital component of culture media that serves as a major energy source for cells in culture. As a component in oral supplementation, it is useful for patients who have advanced stages of cancer to reverse cachexia (i.e. muscle wasting). It is also used for attention deficit-hyperactivity disorder (ADHD), a urinary condition called cystinuria, sickle cell anemia and for alcohol withdrawal support. It finds applications in patients with HIV (AIDS) to prevent weight loss.
Solubility
Soluble in water, dimethyl sulfoxide and ethanol. Insoluble in methanol, ether, benzene, acetone, ethyl acetate and chloroform.
Notes
Moisture and light sensitive. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with moisture and strong oxidizing agents.
Chemical Identifiers
56-85-9 | |
146.15 | |
ZDXPYRJPNDTMRX-VKHMYHEASA-N | |
5961 | |
N[C@@H](CCC(N)=O)C(O)=O |
C5H10N2O3 | |
MFCD00008044 | |
l-glutamine, glutamine, levoglutamide, l-+-glutamine, glutamic acid amide, h-gln-oh, stimulina, cebrogen, glumin, levoglutamid | |
CHEBI:18050 |
Specifications
185°C (decomposition) | |
99% | |
MFCD00008044 | |
l-glutamine, glutamine, levoglutamide, l-+-glutamine, glutamic acid amide, h-gln-oh, stimulina, cebrogen, glumin, levoglutamid | |
ZDXPYRJPNDTMRX-VKHMYHEASA-N | |
(2S)-2-amino-4-carbamoylbutanoic acid | |
5961 | |
146.15 |
56-85-9 | |
C5H10N2O3 | |
1723797 | |
Soluble in water,dimethyl sulfoxide and ethanol. Insoluble in methanol,ether,benzene,acetone,ethyl acetate and chloroform. | |
N[C@@H](CCC(N)=O)C(O)=O | |
146.15 | |
CHEBI:18050 | |
L-Glutamine |
Safety and Handling
P261-P264b-P271-P280g-P280i-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H320-H335-H336
missing translation for 'einecsNumber' : 200-292-1
missing translation for 'rtecsNumber' : MA2275100
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only