missing translation for 'onlineSavingsMsg'
Learn More

L-Histidine hydrochloride monohydrate, 98%, Thermo Scientific Chemicals

40.70 € - 1901.46 €

Chemical Identifiers

CAS 5934-29-2
Molecular Formula C6H12ClN3O3
Molecular Weight (g/mol) 209.63
MDL Number MFCD00151027
InChI Key CMXXUDSWGMGYLZ-XRIGFGBMSA-N
Synonym l-histidine hydrochloride hydrate, l-histidine hydrochloride monohydrate, s-2-amino-3-1h-imidazol-4-yl propanoic acid hydrochloride hydrate, h-his-oh.hcl.h2o, l-histidine monohydrochloride monohydrate, l-histidine, monohydrochloride, monohydrate, l-histidin hydrate hydrochloride, histidine, monohydrochloride, monohydrate, l, h-his-ohhclh2o, l-histidine, hydrochloride, monohydrate
PubChem CID 165377
IUPAC Name (2S)-2-amino-3-(1H-imidazol-5-yl)propanoic acid;hydrate;hydrochloride
SMILES C1=C(NC=N1)CC(C(=O)O)N.O.Cl
View More Specs

Products 4
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10761442
View Documents
Thermo Scientific Acros
411730250
25 g Plastic Bottle
40.70 €
25g
Please to purchase this item. Need a web account? Register with us today!
10095790
View Documents
Thermo Scientific Acros
411731000
100 g Plastic Bottle
115.58 €
100g
Please to purchase this item. Need a web account? Register with us today!
10444045
View Documents
Thermo Scientific Acros
411735000
500 g Plastic Bottle
438.18 €
500g
Please to purchase this item. Need a web account? Register with us today!
10277563
View Documents
Thermo Scientific Acros
411730025
2.5 kg Plastic Bottle
1901.46 €
2.50kg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

5934-29-2
209.63
CMXXUDSWGMGYLZ-XRIGFGBMSA-N
165377
C1=C(NC=N1)CC(C(=O)O)N.O.Cl
C6H12ClN3O3
MFCD00151027
l-histidine hydrochloride hydrate, l-histidine hydrochloride monohydrate, s-2-amino-3-1h-imidazol-4-yl propanoic acid hydrochloride hydrate, h-his-oh.hcl.h2o, l-histidine monohydrochloride monohydrate, l-histidine, monohydrochloride, monohydrate, l-histidin hydrate hydrochloride, histidine, monohydrochloride, monohydrate, l, h-his-ohhclh2o, l-histidine, hydrochloride, monohydrate
(2S)-2-amino-3-(1H-imidazol-5-yl)propanoic acid;hydrate;hydrochloride

Specifications

200ppm max.
3 to 5 (2.5% soln. at 25°C)
10ppm max.
10ppm max.
7% to 10% (3 hrs) (145 to 150°C)
C6H12ClN3O3
25, 513
300ppm max.
0.1% max.
CMXXUDSWGMGYLZ-XRIGFGBMSA-N
(2S)-2-amino-3-(1H-imidazol-5-yl)propanoic acid;hydrate;hydrochloride
209.63
209.64
Crystalline Powder or Crystals
5934-29-2
White
Authentic
98%
0.5% max. (TLC)
MFCD00151027
+ 9.90
10, 4617
l-histidine hydrochloride hydrate, l-histidine hydrochloride monohydrate, s-2-amino-3-1h-imidazol-4-yl propanoic acid hydrochloride hydrate, h-his-oh.hcl.h2o, l-histidine monohydrochloride monohydrate, l-histidine, monohydrochloride, monohydrate, l-histidin hydrate hydrochloride, histidine, monohydrochloride, monohydrate, l, h-his-ohhclh2o, l-histidine, hydrochloride, monohydrate
C1=C(NC=N1)CC(C(=O)O)N.O.Cl
+ 9.90 (20.00°C c=11,6 N HCl)
165377
98%
L-Histidine hydrochloride monohydrate, 98%
Videos
Safety and Handling

Safety and Handling

missing translation for 'rtecsNumber' : MS3119000

Product Identifier
  • L-Histidine hydrochloride monohydrate

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only