missing translation for 'onlineSavingsMsg'
Learn More
L-Histidine methyl ester dihydrochloride, 98+%, Thermo Scientific Chemicals Product Code.: 11419543

L-Histidine methyl ester dihydrochloride, 98+%, Thermo Scientific Chemicals

Product Code. 11419543
5 g
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy

Product Code. 11419543

Brand: Thermo Scientific Alfa Aesar A12386.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

L-Histidine methyl ester dihydrochloride is used to prepare optically pure L-(+)-ergothioneine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
L-Histidine methyl ester dihydrochloride is used to prepare optically pure L-(+)-ergothioneine.

Solubility
Soluble in dimethyl sulfoxide, methanol and water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents. It is hygroscopic in nature. Store under dry inert gas.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7389-87-9
Molecular Formula C7H11ClN3O2
Molecular Weight (g/mol) 204.63
MDL Number MFCD00012701
InChI Key VXXIPPPPKNWFLK-UHFFFAOYNA-N
Synonym l-histidine methyl ester dihydrochloride, h-his-ome.2hcl, methyl l-histidinate dihydrochloride, l-histidine, methyl ester, dihydrochloride, s-methyl 2-amino-3-1h-imidazol-4-yl propanoate dihydrochloride, l-+-histidine methyl ester dihydrochloride, methyl 2s-2-amino-3-1h-imidazol-4-yl propanoate dihydrochloride, methyl 2s-2-amino-3-3h-imidazol-4-yl propanoate dihydrochloride, histidine methyl ester dihydrochloride, c7h11n3o2.2hclShow MoreShow Less
PubChem CID 2723645
SMILES [Cl].COC(=O)C(N)CC1=CN=CN1

Specifications

Melting Point 198°C (decomposition)
Beilstein 3572010
Solubility Information Soluble in dimethyl sulfoxide,methanol and water.
IUPAC Name methyl 2-amino-3-(1H-imidazol-5-yl)propanoate hydrochloridyl
Quantity 5 g
Formula Weight 242.11
Optical Rotation +10° (c=2 in water)
Sensitivity Hygroscopic
Percent Purity ≥98%
Chemical Name or Material L-Histidine methyl ester dihydrochloride

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10244289 Certificate TypeCertificate of Analysis Date26/10/2022 Catalog NumberA12386.06,A12386.14,A12386.22
Lot Number10237703 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog NumberA12386.06,A12386.14,A12386.22
Lot Number10140646 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA12386.06,A12386.14,A12386.22
Lot Number10209835 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA12386.06,A12386.14,A12386.22
Lot Number10209829 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA12386.06,A12386.14,A12386.22

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
L-Histidine methyl ester dihydrochloride, 98+%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.