missing translation for 'onlineSavingsMsg'
Learn More
Thermo Scientific Chemicals L-Histidine monohydrochloride monohydrate, 99% Product Code.: 11408267

Thermo Scientific Chemicals L-Histidine monohydrochloride monohydrate, 99%

Product Code. 11408267
50 g
Click to view available options
Quantity:
50 g
250 g
500 g
This item is not returnable. View return policy

Product Code. 11408267

Brand: Thermo Scientific Chemicals A17627.18

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

L-Histidine monohydrochloride monohydrate, CAS # 5934-29-2, is a histidine precursor through histidine decarboxylase.

L-Histidine monohydrochloride monohydrate is used as a precursor through histidine decarboxylase. It is also used to study the cultures of the human T-lymphoblastic leukemia cell line, modulation of apoptosis. It is also used to enhance the biosynthesis of Lovastatin by cultured aspergillus terreus. It is utilized as a single nitrogen source to probe swarming in Pseudomonas aeruginosa on agar. It is employed as a component to avoid the risk of contaminating bioproducts with adventitious viruses.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

• L-Histidine monohydrochloride monohydrate is a histidine precursor via histidine decarboxylase.

Application

• As a precursor of histidine, it can be used to enhance the biosyntyhesis of lovastatin by cultured Aspergillus terreus
• It is utilized as a single nitrogen source to probe swarming in Pseudomonas aeruginosa on agar
• It can be employed in laboraotry applications as a component to avoid the risk of contaminating bioproducts with adventitious viruses
• It is also used for laboratory study of cultures of the human T-lymphoblastic leukemia cell line, and modulation of apoptosis

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5934-29-2
Molecular Formula C6H12ClN3O3
Molecular Weight (g/mol) 209.63
MDL Number MFCD00151027
InChI Key CMXXUDSWGMGYLZ-XRIGFGBMSA-N
Synonym l-histidine hydrochloride hydrate, l-histidine hydrochloride monohydrate, s-2-amino-3-1h-imidazol-4-yl propanoic acid hydrochloride hydrate, h-his-oh.hcl.h2o, l-histidine monohydrochloride monohydrate, l-histidine, monohydrochloride, monohydrate, l-histidin hydrate hydrochloride, histidine, monohydrochloride, monohydrate, l, h-his-ohhclh2o, l-histidine, hydrochloride, monohydrate
PubChem CID 165377
IUPAC Name (2S)-2-amino-3-(1H-imidazol-4-yl)propanoic acid hydrate hydrochloride
SMILES O.Cl.N[C@@H](CC1=CNC=N1)C(O)=O

Specifications

Melting Point ∽240°C (decomposition)
Assay Percent Range 99%
Beilstein 4168261
Merck Index 14,4720
Solubility Information Soluble in water.
Quantity 50 g
Formula Weight 209.63 (191.62 anhydrous)
Percent Purity 99%
Chemical Name or Material L-Histidine Monohydrochloride Monohydrate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific Chemicals L-Histidine monohydrochloride monohydrate, 99% >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.