missing translation for 'onlineSavingsMsg'
Learn More

L-Histidine, Multi-Compendial, U.S.P., J.T. Baker™

Product Code. 13274004
Click to view available options
Quantity:
100 g
1 kg
Packaging:
Poly Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 71-00-1
Molecular Formula C6H9N3O2
Molecular Weight (g/mol) 155.16
MDL Number MFCD00064315
InChI Key HNDVDQJCIGZPNO-UHFFFAOYNA-N
Synonym l-histidine, histidine, h-his-oh, glyoxaline-5-alanine, anti-rheuma, l---histidine, istidina, s-histidine, l-histidin
PubChem CID 6274
ChEBI CHEBI:15971
IUPAC Name 2-amino-3-(1H-imidazol-5-yl)propanoic acid
SMILES NC(CC1=CN=CN1)C(O)=O

Specifications

Packaging Poly Bottle
Formula Weight 155.16
Grade Multi-Compendial/USP
Quantity 1 kg
Specific Gravity 1L = 1.44kg
Chemical Name or Material L-Histidine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'