missing translation for 'onlineSavingsMsg'
Learn More

L-Histidine, Multi-Compendial, U.S.P., J.T. Baker™

315.00 € - 2690.00 €

Chemical Identifiers

CAS 71-00-1
Molecular Formula C6H9N3O2
Molecular Weight (g/mol) 155.16
MDL Number MFCD00064315
InChI Key HNDVDQJCIGZPNO-UHFFFAOYNA-N
Synonym l-histidine, histidine, h-his-oh, glyoxaline-5-alanine, anti-rheuma, l---histidine, istidina, s-histidine, l-histidin
PubChem CID 6274
ChEBI CHEBI:15971
IUPAC Name 2-amino-3-(1H-imidazol-5-yl)propanoic acid
SMILES NC(CC1=CN=CN1)C(O)=O
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
15517964
View Documents
Avantor J.T.Baker
2080-05
100 g Poly Bottle This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
13274004
View Documents
Avantor J.T.Baker
2080-06
1 kg Poly Bottle
2690.00 €
1kg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Chemical Identifiers

71-00-1
155.16
HNDVDQJCIGZPNO-UHFFFAOYNA-N
6274
2-amino-3-(1H-imidazol-5-yl)propanoic acid
C6H9N3O2
MFCD00064315
l-histidine, histidine, h-his-oh, glyoxaline-5-alanine, anti-rheuma, l---histidine, istidina, s-histidine, l-histidin
CHEBI:15971
NC(CC1=CN=CN1)C(O)=O

Specifications

71-00-1
MFCD00064315
HNDVDQJCIGZPNO-UHFFFAOYNA-N
2-amino-3-(1H-imidazol-5-yl)propanoic acid
6274
155.16
1L = 1.44kg
C6H9N3O2
l-histidine, histidine, h-his-oh, glyoxaline-5-alanine, anti-rheuma, l---histidine, istidina, s-histidine, l-histidin
NC(CC1=CN=CN1)C(O)=O
155.16
CHEBI:15971
Multi-Compendial/USP
L-Histidine
Videos
SDS
Documents

Documents

Product Certifications