missing translation for 'onlineSavingsMsg'
Learn More
Thermo Scientific Chemicals L-Isoleucine, 99% Product Code.: 10563721

Thermo Scientific Chemicals L-Isoleucine, 99%

Product Code. 10563721
25 g, Plastic Bottle
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
Packaging:
Glass Bottle
Plastic Bottle
This item is not returnable. View return policy

Product Code. 10563721

Brand: Thermo Scientific Chemicals 166170250

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 73-32-5
Molecular Formula C6H13NO2
Molecular Weight (g/mol) 131.18
MDL Number MFCD00064222 MFCD00004268
InChI Key AGPKZVBTJJNPAG-WHFBIAKZSA-N
Synonym l-isoleucine, isoleucine, 2s,3s-2-amino-3-methylpentanoic acid, s-isoleucine, s,s-isoleucine, 2s,3s-isoleucine, 2-amino-3-methylvaleric acid, erythro-l-isoleucine, l-+-isoleucine, l-ileShow MoreShow Less
PubChem CID 6306
ChEBI CHEBI:17191
IUPAC Name (2S,3S)-2-amino-3-methylpentanoic acid
SMILES CC[C@H](C)[C@H](N)C(O)=O

Specifications

Melting Point 284.0°C to 286.0°C
Color White
Chloride 500ppm max.
Heavy Metals (as Pb) 20ppm max.
Infrared Spectrum Authentic
Iron (Fe) 30ppm max.
Assay Percent Range 99%
Loss on Drying 0.3% max. (105°C, 3 hrs)
Packaging Plastic Bottle
Linear Formula C2H5CH(CH3)CH(NH2)CO2H
Residue after Ignition 0.3% max.
Beilstein 04, 454
Specific Rotation + 41.00
Sulfate 300ppm max.
Merck Index 15, 5225
Solubility Information Solubility in water: 41.2g/L (50°C). Other solubilities: soluble in mineral acids, practically insoluble in alcohol, insoluble in ether
Sublimation Point 168.0 °C
Specific Rotation Condition +41° (20°C c=4,6N HCl)
Quantity 25 g
Formula Weight 131.17
Percent Purity 99%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material L-Isoleucine
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0466613 Certificate TypeCertificate of Analysis Date07/10/2024 Catalog Number166170250,166171000,166175000,166170050
Lot NumberA0461657 Certificate TypeCertificate of Analysis Date11/06/2024 Catalog Number166170250,166171000,166175000,166170050
Lot NumberA0451304 Certificate TypeCertificate of Analysis Date02/05/2023 Catalog Number166170250,166171000,166175000,166170050
Lot NumberA0398921 Certificate TypeCertificate of Analysis Date15/12/2022 Catalog Number166170250,166171000,166175000,166170050
Lot NumberA0396942 Certificate TypeCertificate of Analysis Date15/12/2022 Catalog Number166170250,166171000,166175000,166170050

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific Chemicals L-Isoleucine, 99% >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.