missing translation for 'onlineSavingsMsg'
Learn More
Thermo Scientific Chemicals L-Isoleucine, 99% Product Code.: 11484684

Thermo Scientific Chemicals L-Isoleucine, 99%

Product Code. 11484684
10 g
Click to view available options
Quantity:
10 g
50 g
100 g
250 g
This item is not returnable. View return policy

Product Code. 11484684

Brand: Thermo Scientific Chemicals A13699.09

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

L-isoleucine, CAS # 73-32-5, is a branched-chain aliphatic amino acid that is an essential constituent of the diet and is also used in several biological mechanisms, including the biosynthesis of proteins.

L-Isoleucine acts as an essential amino acid in human. It is used as a flavoring agent and nutritional supplement for foods. It is also used as an antipsychotic. It is employed as a component of proteins. Further, it stimulates immune function and promotes secretion of several hormones. In addition to this, it is used for hemoglobin formation and regulating blood sugar and energy levels.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

• L-isoleucine is an essential amino acid that organisms are incapable of synthesizing. It must be ingested in the diet and can be found mainly in meats, fish, cheese, eggs, seeds, and nuts
• L-Isoleucine is an isomer of L-leucine. It is synthesized from threonine and is a branched-chain hydrophobic amino acid

Application

• L-isoleucine is used in several biological mechanisms, including assisting wound healing, detoxifying nitrogenous wastes, stimulating immune function, and promoting hormone secretion
• L-isoleucine shows antipsychotic activity
• It is important for hemoglobin formation and regulating blood sugar and energy levels
• L-isoleucine participates in protein synthesis by binding with transfer RNA (tRNA) in the cell cytoplasm
• This compound also plays an important role as a metabolite for several species, including Saccharomyces cerevisiae, Escherichia coli, plants, algae, and mice
• L-isoleucine can be used as an ingredient for the development of other essential biochemical components in the organism, some of which are used for energy production and stimulants in the upper brain
• It has been used as a supplement of maintenance media (DMEM/F-12) for the culture of primary hepatocytes
• It has been used in capillary feeder assay to determine the consumption of amino acids by Drosophila melanogaster

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 73-32-5
Molecular Formula C6H13NO2
Molecular Weight (g/mol) 131.18
MDL Number MFCD00064222 MFCD00004268
InChI Key AGPKZVBTJJNPAG-WHFBIAKZSA-N
Synonym l-isoleucine, isoleucine, 2s,3s-2-amino-3-methylpentanoic acid, s-isoleucine, s,s-isoleucine, 2s,3s-isoleucine, 2-amino-3-methylvaleric acid, erythro-l-isoleucine, l-+-isoleucine, l-ileShow MoreShow Less
PubChem CID 6306
ChEBI CHEBI:17191
IUPAC Name (2S,3S)-2-amino-3-methylpentanoic acid
SMILES CC[C@H](C)[C@H](N)C(O)=O

Specifications

Melting Point ∽287°C (decomposition)
Assay Percent Range 99%
Beilstein 1721792
Merck Index 14,5179
Solubility Information Soluble in water. Slightly soluble in hot alcohol and hot acetic acid. Insoluble in ether.
Quantity 10 g
Formula Weight 131.18
Percent Purity 99%
Chemical Name or Material L-Isoleucine

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10158689 Certificate TypeCertificate of Analysis Date30/08/2024 Catalog NumberA13699.22,A13699.30,A13699.18,A13699.09
Lot NumberQ18J002 Certificate TypeCertificate of Analysis Date24/04/2023 Catalog NumberA13699.09
Lot NumberU08I025 Certificate TypeCertificate of Analysis Date12/08/2022 Catalog NumberA13699.22,A13699.30,A13699.18,A13699.09
Lot NumberS24I039 Certificate TypeCertificate of Analysis Date02/08/2022 Catalog NumberA13699.22,A13699.30,A13699.18,A13699.09
Lot NumberY20E002 Certificate TypeCertificate of Analysis Date27/07/2022 Catalog NumberA13699.22,A13699.30,A13699.18,A13699.09

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific Chemicals L-Isoleucine, 99% >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.