missing translation for 'onlineSavingsMsg'
Learn More
Thermo Scientific Chemicals L-Isoleucine, Cell Culture Reagent Product Code.: 15435319

Thermo Scientific Chemicals L-Isoleucine, Cell Culture Reagent

Product Code. 15435319
100 g
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy

Product Code. 15435319

Brand: Thermo Scientific Chemicals J63045.22

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 73-32-5
Molecular Formula C6H13NO2
Molecular Weight (g/mol) 131.18
MDL Number MFCD00064222 MFCD00004268
InChI Key AGPKZVBTJJNPAG-WHFBIAKZSA-N
Synonym l-isoleucine, isoleucine, 2s,3s-2-amino-3-methylpentanoic acid, s-isoleucine, s,s-isoleucine, 2s,3s-isoleucine, 2-amino-3-methylvaleric acid, erythro-l-isoleucine, l-+-isoleucine, l-ile
PubChem CID 6306
ChEBI CHEBI:17191
IUPAC Name (2S,3S)-2-amino-3-methylpentanoic acid
SMILES CC[C@H](C)[C@H](N)C(O)=O

Specifications

Melting Point ∽287°C (decomposition)
Color White
Beilstein 1721792
Merck Index 14,5179
Quantity 100 g
Formula Weight 131.18
Optical Rotation +40° (c=5 in 6N HCl)
Grade Cell Culture Reagent
Physical Form Powder
Chemical Name or Material L-Isoleucine

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberM21L035 Certificate TypeCertificate of Analysis Date28/01/2025 Catalog NumberJ63045.14,J63045.22
Lot NumberM13L012 Certificate TypeCertificate of Analysis Date14/01/2025 Catalog NumberJ63045.14,J63045.22
Lot NumberZ05J025 Certificate TypeCertificate of Analysis Date05/01/2024 Catalog NumberJ63045.14,J63045.22
Lot NumberY15I003 Certificate TypeCertificate of Analysis Date22/11/2022 Catalog NumberJ63045.14,J63045.22
Lot NumberS13I010 Certificate TypeCertificate of Analysis Date22/06/2022 Catalog NumberJ63045.14,J63045.22

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific Chemicals L-Isoleucine, Cell Culture Reagent >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.