Learn More
L-Isoleucine methyl ester hydrochloride, 98+%, Thermo Scientific Chemicals
Description
L-Isoleucine methyl ester used for chemical synthesis. E.g. It can also be used to produce 3-methyl-2-(4-nitro-imidazol-1-yl)-pentanoic acid methyl ester. It also finds its use as a pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
L-Isoleucine methyl ester used for chemical synthesis. E.g. It can also be used to produce 3-methyl-2-(4-nitro-imidazol-1-yl)-pentanoic acid methyl ester. It also finds its use as a pharmaceutical intermediate.
Solubility
Soluble in methanol (50 mg/ml clear, colorless solution).
Notes
Hygroscopic. Keep away from oxidizing agents, water/moisture. Store under inert atmosphere. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. In order to obtain a higher solubility, warm the tube at 37°C and shake it in the ultrasonic bath for a while.

Specifications
Specifications
Melting Point | 98°C to 100°C |
Beilstein | 3595132 |
Solubility Information | Soluble in methanol (50mg/ml clear,colorless solution). |
Quantity | 25 g |
Formula Weight | 181.66 |
Sensitivity | Hygroscopic |
Percent Purity | ≥98% |
Chemical Name or Material | L-Isoleucine methyl ester hydrochloride |
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
10237162 | Certificate of Analysis | 23/08/2024 | A17847.06,A17847.03,A17847.14 |
10224378 | Certificate of Analysis | 23/08/2024 | A17847.06,A17847.03,A17847.14 |
S26E041 | Certificate of Analysis | 19/07/2022 | A17847.06,A17847.03,A17847.14 |
I2183A | Certificate of Analysis | 23/11/2021 | A17847.06,A17847.03,A17847.14 |
10237162 | Certificate of Analysis | 23/11/2021 | A17847.06,A17847.03,A17847.14 |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.