missing translation for 'onlineSavingsMsg'
Learn More
L-Isoleucine, Multi-Compendial, U.S.P., J.T. Baker™ Product Code.: 15537964

L-Isoleucine, Multi-Compendial, U.S.P., J.T. Baker™

Product Code. 15537964
1 kg, Poly Bottle
Click to view available options
Quantity:
100 g
1 kg
Packaging:
Poly Bottle
This item is not returnable. View return policy

Product Code. 15537964

Brand: Avantor J.T.Baker 208206

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 73-32-5
Molecular Formula C6H13NO2
Molecular Weight (g/mol) 131.18
MDL Number MFCD00064222 MFCD00004268
InChI Key AGPKZVBTJJNPAG-WHFBIAKZSA-N
Synonym l-isoleucine, isoleucine, 2s,3s-2-amino-3-methylpentanoic acid, s-isoleucine, s,s-isoleucine, 2s,3s-isoleucine, 2-amino-3-methylvaleric acid, erythro-l-isoleucine, l-+-isoleucine, l-ile
PubChem CID 6306
ChEBI CHEBI:17191
IUPAC Name (2S,3S)-2-amino-3-methylpentanoic acid
SMILES CC[C@H](C)[C@H](N)C(O)=O

Specifications

Packaging Poly Bottle
Quantity 1 kg
Formula Weight 131.17
Grade Multi-Compendial/USP
Chemical Name or Material L-Isoleucine
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
L-Isoleucine, Multi-Compendial, U.S.P., J.T. Baker™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.