Learn More
L-Isoleucine tert-butyl ester hydrochloride, 98%, Thermo Scientific Chemicals
Description
It is used as a pharmaceutical industry. t is an important raw material and intermediate used in organic synthesis, agrochemicals and dyestuffs.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
It is used as a pharmaceutical industry. t is an important raw material and intermediate used in organic synthesis, agrochemicals and dyestuffs.
Solubility
Soluble in 50 mg/ml methanol.
Notes
Air sensitive. Incompatible with strong oxidizing agents.

Specifications
Specifications
Linear Formula | C10H21NO2·HCl |
Solubility Information | Soluble in 50mg/ml methanol. |
Quantity | 5 g |
Formula Weight | 223.74 |
Percent Purity | 98% |
Physical Form | Solid |
Chemical Name or Material | L-Isoleucine tert-butyl ester hydrochloride |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.