missing translation for 'onlineSavingsMsg'
Learn More

L-Isoleucine (White Crystals or Crystalline Powder), Fisher BioReagents™

Product Code. 10153293
100 g
Click to view available options
Quantity:
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy
This amino acid is suitable for use in tissue culture systems requiring additives.
TRUSTED_SUSTAINABILITY

Specifications

Melting Point 284°C
Ammonium 0.02% max.
CAS 73-32-5
Arsenic (As) 1ppm max.
pH 5.5 to 6.5
Color White
Heavy Metals (as Pb) 10ppm max.
Ignition Residue 0.1% max.
Identification Pass Test
Assay Percent Range 98.5 to 101.0 %
Loss on Drying 0.2% max.
Other Amino Acids Chromatographically not detectable
Molecular Formula C6H13NO2
Packaging Glass Bottle
MDL Number MFCD00064222 MFCD00004268
Specific Rotation 39.5 to 41.5° (+ or -)
Merck Index 15, 5225
State of Solution (%T) 98.0% min.
Synonym l-isoleucine, isoleucine, 2s,3s-2-amino-3-methylpentanoic acid, s-isoleucine, s,s-isoleucine, 2s,3s-isoleucine, 2-amino-3-methylvaleric acid, erythro-l-isoleucine, l-+-isoleucine, l-ile
Alpha Vector Isoleucine
InChI Key AGPKZVBTJJNPAG-WHFBIAKZSA-N
SMILES CC[C@H](C)[C@H](N)C(O)=O
IUPAC Name (2S,3S)-2-amino-3-methylpentanoic acid
Molecular Weight (g/mol) 131.18
Quantity 100 g
PubChem CID 6306
ChEBI CHEBI:17191
Formula Weight 131.17
Percent Purity 98.5 to 101.0%
Physical Form Powder
Chemical Name or Material L-Isoleucine
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
L-Isoleucine (White Crystals or Crystalline Powder), Fisher BioReagents™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.