missing translation for 'onlineSavingsMsg'
Learn More

L(+)-Lactic acid, 20 wt.% solution in water

Product Code. 10522331 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 L
2.5 L
Packaging:
Glass bottle
Unit Size:
1 L
2.5 L
This item is not returnable. View return policy

Product Code. 10522331

Brand: Thermo Scientific Acros undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 79-33-4 | C3H6O3 | 90.078 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 79-33-4, 7732-18-5
Molecular Formula C3H6O3
Molecular Weight (g/mol) 90.078
MDL Number MFCD00064266
InChI Key JVTAAEKCZFNVCJ-REOHCLBHSA-N
Synonym l-lactic acid, l-+-lactic acid, s-2-hydroxypropanoic acid, s-lactic acid, sarcolactic acid, s-2-hydroxypropionic acid, 2s-2-hydroxypropanoic acid, +-lactic acid, espiritin, tisulac
PubChem CID 107689
ChEBI CHEBI:422
IUPAC Name (2S)-2-hydroxypropanoic acid
SMILES CC(C(=O)O)O

Specifications

Color Colorless
Beilstein 03, IV, 633
Fieser 15,181
Merck Index 15, 5385
Formula Weight 90.08
Percent Purity ≥18%
Density 1.0500g/mL
Quantity 1 L
Packaging Glass bottle
Physical Form Liquid
Chemical Name or Material L(+)-Lactic acid
Show More Show Less
compliance-icons
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 C
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • EUH071-Corrosive to the respiratory tract.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.