missing translation for 'onlineSavingsMsg'
Learn More

Melford L-Lysine Acetate

L-Lysine Acetate is employed in biotechnological processes, especially in the production of recombinant proteins and enzymes.

137.00 € - 580.00 €

Specifications

Chemical Name or Material L-Lysine Acetate
CAS 57282-49-2
Molecular Formula C8H18N2O4
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30018435
View Documents
Melford
L37020-100.0
100 g
137.00 €
100g
Please to purchase this item. Need a web account? Register with us today!
30018436
View Documents
Melford
L37020-500.0
500 g
580.00 €
500g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

L-Lysine Acetate is employed in biotechnological processes, especially in the production of recombinant proteins and enzymes.

L-Lysine Acetate is employed in biotechnological processes, especially in the production of recombinant proteins and enzymes. It plays a role in cell culture media to support the growth and productivity of cells.

Additionally, L-Lysine Acetate is utilized in laboratory research, particularly in studies related to protein synthesis, enzymology, and other biochemical processes.

Its role as a fundamental amino acid makes it valuable in understanding various cellular and molecular mechanisms.

Specifications

Specifications

L-Lysine Acetate
57282-49-2
C8H18N2O4
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Melford L-Lysine Acetate

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.