missing translation for 'onlineSavingsMsg'
Learn More
Thermo Scientific Chemicals L(+)-Lysine monohydrate, 99% Product Code.: 10625042

Thermo Scientific Chemicals L(+)-Lysine monohydrate, 99%

Product Code. 10625042
25 g, Plastic Bottle
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Plastic Bottle
This item is not returnable. View return policy

Product Code. 10625042

Brand: Thermo Scientific Chemicals 173730250

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 39665-12-8
Molecular Formula C6H16N2O3
Molecular Weight (g/mol) 164.21
MDL Number MFCD00151035
InChI Key HZRUTVAFDWTKGD-XZNNNFJINA-N
Synonym l-lysine monohydrate, l-lysine hydrate, lysine monohydrate, l +-lysine monohydrate, l-lysine, hydrate, h-lys-oh h2o, lysine, hydrate, lysine hydrate, l-lysine, monohydrate, l-lysine, hydrate 1:1
PubChem CID 16211825
SMILES O.NCCCC[C@H](N)C(O)=O

Specifications

Melting Point 212.0°C to 214.0°C
Ammonium 200ppm max.
pH 9 to 10.5 (2.5% soln. at 25°C)
Color White to Yellow
Chlorine (Cl) 300ppm max.
Heavy Metals (as Pb) 10ppm max.
Infrared Spectrum Authentic
Iron (Fe) 10ppm max.
Assay Percent Range 99%
Other Amino Acids 0.5% max. (TLC)
Packaging Plastic Bottle
Linear Formula H2N(CH2)4CH(NH2)CO2H·H2O
Beilstein 04, 435
Specific Rotation + 26.25
Sulfate 200ppm max.
Merck Index 14, 5636
Transmittance 95% min. (c=5, H2O, 430nm) 1cm cell
Sulfated Ash 0.1% max.
Solubility Information Solubility in water: 1500g/L (20°C). Other solubilities: insoluble in common neutral solvents
Water 10 to 11.5%
IUPAC Name (2S)-2,6-diaminohexanoic acid hydrate
Specific Rotation Condition + 26.25 (20.00°C c=8,6N HCl)
Quantity 25 g
Formula Weight 164.21
Percent Purity 99%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material L(+)-Lysine monohydrate
Show More Show Less
Product Identifier
  • L(+)-Lysine monohydrate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific Chemicals L(+)-Lysine monohydrate, 99% >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.