missing translation for 'onlineSavingsMsg'
Learn More
Thermo Scientific Chemicals L-Lysine monohydrochloride, Cell Culture Reagent Product Code.: 15430227

Thermo Scientific Chemicals L-Lysine monohydrochloride, Cell Culture Reagent

Product Code. 15430227
100 g
missing translation for 'orderingAttributeHoverText'
Quantity:
100 g
500 g
1 kg
This item is not returnable. View return policy

Product Code. 15430227

Brand: Thermo Scientific Chemicals J62099.22

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 657-27-2
Molecular Formula C6H15ClN2O2
Molecular Weight (g/mol) 182.65
MDL Number MFCD00064564 MFCD00081870
InChI Key BVHLGVCQOALMSV-JEDNCBNOSA-N
Synonym l-lysine hydrochloride, l-lysine monohydrochloride, lysine hydrochloride, h-lys-oh.hcl, l-lysine, monohydrochloride, lyamine, darvyl, lysion, s-2,6-diaminohexanoic acid hydrochloride, enisyl
PubChem CID 69568
ChEBI CHEBI:53633
IUPAC Name (2S)-2,6-diaminohexanoic acid;hydrochloride
SMILES Cl.NCCCC[C@H](N)C(O)=O

Specifications

Color White
Beilstein 3563889
Merck Index 14,5636
Quantity 100 g
Formula Weight 182.65
Grade Cell Culture Reagent
Physical Form Powder
Chemical Name or Material L-Lysine monohydrochloride

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific Chemicals L-Lysine monohydrochloride, Cell Culture Reagent >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.