missing translation for 'onlineSavingsMsg'
Learn More
L-(-)-Malic acid, 97%, Thermo Scientific Chemicals Product Code.: 11428860

L-(-)-Malic acid, 97%, Thermo Scientific Chemicals

Product Code. 11428860
250 g
Click to view available options
Quantity:
100 g
250 g
500 g
This item is not returnable. View return policy

Product Code. 11428860

Brand: Thermo Scientific Alfa Aesar A13702.30

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

L-(-)-Malic acid is used as Selective α-amino protecting reagent for amino acid derivatives. Versatile synthon for the preparation of chiral compounds including κ-opioid rece. It also acts as active ingredient in many sour or tart foods. It is used as synthesizing disincrustant and fluorescent whitening agent. It aids in the production of polyester and alcohol acid resins.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
L-(-)-Malic acid is used as Selective α-amino protecting reagent for amino acid derivatives. Versatile synthon for the preparation of chiral compounds including κ-opioid rece. It also acts as active ingredient in many sour or tart foods. It is used as synthesizing disincrustant and fluorescent whitening agent. It aids in the production of polyester and alcohol acid resins.

Solubility
Soluble in acetone, dioxane, water, methanol and ethanol. Insoluble in benzene

Notes
Incompatible with Bases, Oxidizing agents, Reducing agents, Alkali metals .
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 97-67-6
Molecular Formula C4H6O5
Molecular Weight (g/mol) 134.087
MDL Number MFCD00064213
InChI Key BJEPYKJPYRNKOW-REOHCLBHSA-N
Synonym l-malic acid, l---malic acid, s-2-hydroxysuccinic acid, 2s-2-hydroxybutanedioic acid, l--malic acid, apple acid, --malic acid, l-apple acid, s-malic acid, s-2-hydroxybutanedioic acidShow MoreShow Less
PubChem CID 222656
ChEBI CHEBI:30797
IUPAC Name (2S)-2-hydroxybutanedioic acid
SMILES C(C(C(=O)O)O)C(=O)O

Specifications

Melting Point 100°C to 106°C
Assay Percent Range 97%
Beilstein 1723541
Merck Index 14,5707
Solubility Information Soluble in acetone, dioxane, water, methanol and ethanol; Insoluble in benzene
Formula Weight 134.09
Percent Purity 97%
Density 1.6
Quantity 250 g
Flash Point 220°C (428°F)
Optical Rotation −27° (c=5.5 in Pyridine)
Chemical Name or Material L-(-)-Malic acid
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10251180 Certificate TypeCertificate of Analysis Date30/08/2024 Catalog NumberA13702.22,A13702.36,A13702.30
Lot NumberK13K14 Certificate TypeCertificate of Analysis Date07/08/2020 Catalog NumberA13702.22,A13702.36,A13702.30
Lot NumberF4163A Certificate TypeCertificate of Analysis Date07/08/2020 Catalog NumberA13702.22,A13702.36,A13702.30
Lot Number10138956 Certificate TypeCertificate of Analysis Date07/08/2020 Catalog NumberA13702.22,A13702.36,A13702.30
Lot Number10183311 Certificate TypeCertificate of Analysis Date07/08/2020 Catalog NumberA13702.22,A13702.36,A13702.30

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
L-(-)-Malic acid, 97%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.