missing translation for 'onlineSavingsMsg'
Learn More
L(-)-Malic acid, 99%, Thermo Scientific Chemicals Product Code.: 10295290

L(-)-Malic acid, 99%, Thermo Scientific Chemicals

Product Code. 10295290
100 g, Plastic bottle
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Plastic bottle
This item is not returnable. View return policy

Product Code. 10295290

Brand: Thermo Scientific Acros 150591000

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 97-67-6
Molecular Formula C4H6O5
Molecular Weight (g/mol) 134.087
MDL Number MFCD00064213
InChI Key BJEPYKJPYRNKOW-REOHCLBHSA-N
Synonym l-malic acid, l---malic acid, s-2-hydroxysuccinic acid, 2s-2-hydroxybutanedioic acid, l--malic acid, apple acid, --malic acid, l-apple acid, s-malic acid, s-2-hydroxybutanedioic acidShow MoreShow Less
PubChem CID 222656
ChEBI CHEBI:30797
IUPAC Name (2S)-2-hydroxybutanedioic acid
SMILES C(C(C(=O)O)O)C(=O)O

Specifications

Melting Point 99°C to 107°C
Color White
Assay Percent Range 99%
Linear Formula HO2CCH2CH(OH)CO2H
Beilstein 03, 419
Merck Index 15, 5771
Formula Weight 134.09
Percent Purity ≥98.5%
Quantity 100 g
Infrared Spectrum Authentic
Packaging Plastic bottle
Physical Form Crystalline Powder or Granules
Chemical Name or Material L(-)-Malic acid
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0415560 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog Number150590250,150591000,150595000
Lot NumberA0457807 Certificate TypeCertificate of Analysis Date27/11/2023 Catalog Number150590250,150591000,150595000
Lot NumberA0401881 Certificate TypeCertificate of Analysis Date15/12/2022 Catalog Number150590250,150591000,150595000
Lot NumberA0394167 Certificate TypeCertificate of Analysis Date15/12/2022 Catalog Number150590250,150591000,150595000
Lot NumberA0415560 Certificate TypeCertificate of Analysis Date25/07/2022 Catalog Number150590250,150591000,150595000
compliance-icons
Product Identifier
  • L(-)-Malic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contains : L(-)-Malic acid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
L(-)-Malic acid, 99%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.