missing translation for 'onlineSavingsMsg'
Learn More
Thermo Scientific Chemicals L-Methionine sulfoxide Product Code.: 15415179

Thermo Scientific Chemicals L-Methionine sulfoxide

Product Code. 15415179
5 g
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy

Product Code. 15415179

Brand: Thermo Scientific Chemicals J62873.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3226-65-1
Molecular Formula C5H11NO3S
Molecular Weight (g/mol) 165.21
MDL Number MFCD00063093
InChI Key QEFRNWWLZKMPFJ-YGVKFDHGSA-N
Synonym l-methionine sulfoxide, h-met o-oh, methionine s-oxide, methionine sulfoxide, 2s-2-amino-4-methylsulfinyl butanoic acid, l-methionine s-oxide, 2s-2-amino-4-methylsulfinylbutanoic acid, l-2-amino-4-methylsulfinyl butanoic acid, l-methionine-dl-sulfoxide, 2s-2-azanyl-4-methylsulfinyl-butanoic acid
PubChem CID 158980
ChEBI CHEBI:17016
IUPAC Name (2S)-2-amino-4-methylsulfinylbutanoic acid
SMILES CS(=O)CC[C@H](N)C(O)=O

Specifications

Color White
Beilstein 4129939
Quantity 5 g
Formula Weight 165.21
Physical Form Powder
Chemical Name or Material L-Methionine sulfoxide

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberP06J016 Certificate TypeCertificate of Analysis Date09/05/2023 Catalog NumberJ62873.06
Lot NumberN17I011 Certificate TypeCertificate of Analysis Date17/11/2022 Catalog NumberJ62873.06,J62873.03,J62873.14
Lot NumberX03I007 Certificate TypeCertificate of Analysis Date13/10/2022 Catalog NumberJ62873.06,J62873.03,J62873.14
Lot NumberT06I028 Certificate TypeCertificate of Analysis Date27/07/2022 Catalog NumberJ62873.06,J62873.03,J62873.14
Lot NumberQ25E019 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberJ62873.06,J62873.03,J62873.14

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific Chemicals L-Methionine sulfoxide >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.