missing translation for 'onlineSavingsMsg'
Learn More

L-Methioninol, 99+%, Thermo Scientific Chemicals

Product Code. 10269823
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2899-37-8
Molecular Formula C5H13NOS
Molecular Weight (g/mol) 135.23
MDL Number MFCD00004735
InChI Key MIQJGZAEWQQAPN-YFKPBYRVSA-N
Synonym l-methioninol, l---methioninol, h-methioninol, 2s-2-amino-4-methylsulfanyl butan-1-ol, s---methioninol, s-2-amino-4-methylthio butan-1-ol, d-s---methionanol, methioninol, 2s-2-amino-4-methylsulfanylbutan-1-ol, s-2-amino-4-methylsulfanyl-butan-1-ol
PubChem CID 2724404
SMILES CSCC[C@H](N)CO

Spezifikation

Infrared Spectrum Authentic
Color White
Packaging Glass bottle
Specific Rotation -11.5° to -13.5° (20°C, 589nm) (c=1.4, ethanol)
Specific Rotation Condition −12.50° (20°C c=1.4,C2H5OH)
Quantity 5 g
Physical Form Crystalline Low Melting Solid
Assay Percent Range 99+%
IUPAC Name (2S)-2-amino-4-methylsulfanylbutan-1-ol
Formula Weight 135.22
Percent Purity ≥99.0%
Chemical Name or Material L-Methioninol
Mehr anzeigen Weniger anzeigen
compliance-icons
Produktidentifikator
  • L-Methioninol
Signalwort
  • Warning
Gefahrenkategorie
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Gefahrenhinweis
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Sicherheitshinweis
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'