missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L-(+)-Pantolactone, 98%, Thermo Scientific™
112.00 € - 331.00 €
Chemical Identifiers
CAS | 5405-40-3 |
---|---|
Molecular Formula | C6H10O3 |
Molecular Weight (g/mol) | 130.143 |
MDL Number | MFCD00216625 |
InChI Key | SERHXTVXHNVDKA-SCSAIBSYSA-N |
Synonym | s-+-pantolactone, l-pantolactone, s-3-hydroxy-4,4-dimethyldihydrofuran-2 3h-one, s-pantolactone, l-+-pantoyl lactone, 3s-3-hydroxy-4,4-dimethyloxolan-2-one, d---pantolactone, 2 3h-furanone, dihydro-3-hydroxy-4,4-dimethyl-, 3s, pantolactons, l-pantoyl lactone |
PubChem CID | 736053 |
IUPAC Name | (3S)-3-hydroxy-4,4-dimethyloxolan-2-one |
SMILES | CC1(COC(=O)C1O)C |
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
ApplicationsL-(+)-Pantolactone, is used as Pharmaceutical material and intermediate. It is used as an Intermediate in organic syntheses.
Solubility
Soluble in water, hardly soluble in alcohol.
Notes
Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents. Heat sensitive.
Chemical Identifiers
5405-40-3 | |
130.143 | |
SERHXTVXHNVDKA-SCSAIBSYSA-N | |
736053 | |
CC1(COC(=O)C1O)C |
C6H10O3 | |
MFCD00216625 | |
s-+-pantolactone, l-pantolactone, s-3-hydroxy-4,4-dimethyldihydrofuran-2 3h-one, s-pantolactone, l-+-pantoyl lactone, 3s-3-hydroxy-4,4-dimethyloxolan-2-one, d---pantolactone, 2 3h-furanone, dihydro-3-hydroxy-4,4-dimethyl-, 3s, pantolactons, l-pantoyl lactone | |
(3S)-3-hydroxy-4,4-dimethyloxolan-2-one |
Specifications
5405-40-3 | |
120°C to 122°C (15 mmHg) | |
MFCD00216625 | |
s-+-pantolactone, l-pantolactone, s-3-hydroxy-4,4-dimethyldihydrofuran-2 3h-one, s-pantolactone, l-+-pantoyl lactone, 3s-3-hydroxy-4,4-dimethyloxolan-2-one, d---pantolactone, 2 3h-furanone, dihydro-3-hydroxy-4,4-dimethyl-, 3s, pantolactons, l-pantoyl lactone | |
SERHXTVXHNVDKA-SCSAIBSYSA-N | |
CC1(COC(=O)C1O)C | |
130.143 | |
130.14 | |
L-(+)-Pantolactone |
89°C to 94°C | |
C6H10O3 | |
4659425 | |
Soluble in water,hardly soluble in alcohol. | |
+51° (c=3 in Water) | |
(3S)-3-hydroxy-4,4-dimethyloxolan-2-one | |
736053 | |
98% |
Safety and Handling
GHS H Statement
H318
Causes serious eye damage.
P280-P305+P351+P338-P310
H318
missing translation for 'tsca' : No
Recommended Storage : Keep cold
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title