missing translation for 'onlineSavingsMsg'
Learn More
Thermo Scientific Chemicals L-Phenylalanine, Cell Culture Reagent Product Code.: 15406089

Thermo Scientific Chemicals L-Phenylalanine, Cell Culture Reagent

Product Code. 15406089
250 g
Click to view available options
Quantity:
100 g
250 g
500 g
This item is not returnable. View return policy

Product Code. 15406089

Brand: Thermo Scientific Chemicals J63925.30

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

The primary use of L-phenylalanine in the body is in the production of tyrosine, another amino acid. It is an essential amino acid, which is biologically converted into L-tyrosine, another one of the DNA-encoded amino acids, which in turn is converted to L-DOPA and further converted into dopamine, norepinephrine, and epinephrine. L-Phenylalanine is produced for medical, feed, and nutritional applications such as in the preparation of Aspartame.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
The primary use of L-phenylalanine in the body is in the production of tyrosine, another amino acid. It is an essential amino acid, which is biologically converted into L-tyrosine, another one of the DNA-encoded amino acids, which in turn is converted to L-DOPA and further converted into dopamine, norepinephrine, and epinephrine. L-Phenylalanine is produced for medical, feed, and nutritional applications such as in the preparation of Aspartame.

Solubility
Soluble in water. Aqueous solubility increases with warming. Also soluble in methanol, ethanol or 1M NH{4}OH

Notes
Store at room temperature. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 63-91-2
Molecular Formula C9H11NO2
Molecular Weight (g/mol) 165.19
MDL Number MFCD00064227
InChI Key COLNVLDHVKWLRT-UHFFFAOYNA-N
Synonym l-phenylalanine, phenylalanine, s-2-amino-3-phenylpropanoic acid, 3-phenyl-l-alanine, s-phenylalanine, 3-phenylalanine, 2s-2-amino-3-phenylpropanoic acid, s-2-amino-3-phenylpropionic acid, beta-phenyl-l-alanine, h-phe-oh
PubChem CID 6140
ChEBI CHEBI:17295
SMILES NC(CC1=CC=CC=C1)C(O)=O

Specifications

Color White
Beilstein 1910408
Merck Index 14,7271
Solubility Information Soluble in water; Aqueous solubility increases with warming; Also soluble in methanol,ethanol or 1M NH4OH
IUPAC Name (2S)-2-amino-3-phenylpropanoic acid
Quantity 250 g
Formula Weight 165.19
Grade Cell Culture Reagent
Physical Form Powder
Chemical Name or Material L-Phenylalanine

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberW10K037 Certificate TypeCertificate of Analysis Date23/09/2024 Catalog NumberJ63925.30,J63925.22,J63925.36
Lot NumberW21J037 Certificate TypeCertificate of Analysis Date25/10/2023 Catalog NumberJ63925.30,J63925.22,J63925.36
Lot NumberZ09I041 Certificate TypeCertificate of Analysis Date02/02/2023 Catalog NumberJ63925.30,J63925.22,J63925.36
Lot NumberN14I020 Certificate TypeCertificate of Analysis Date17/11/2022 Catalog NumberJ63925.30,J63925.22,J63925.36
Lot NumberX28H060 Certificate TypeCertificate of Analysis Date20/09/2022 Catalog NumberJ63925.30,J63925.22,J63925.36

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific Chemicals L-Phenylalanine, Cell Culture Reagent >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.