missing translation for 'onlineSavingsMsg'
Learn More
L-Phenylalanine methyl ester hydrochloride, 99%, Thermo Scientific Chemicals Product Code.: 11460323

L-Phenylalanine methyl ester hydrochloride, 99%, Thermo Scientific Chemicals

Product Code. 11460323
10 g
Click to view available options
Quantity:
10 g
50 g
250 g
This item is not returnable. View return policy

Product Code. 11460323

Brand: Thermo Scientific Alfa Aesar A11455.09

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Methyl L-phenylalaninate hydrochloride is used in pharmaceutical, food,animal feed,cosmetic,bio-chemical research etc.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl L-phenylalaninate hydrochloride is used in pharmaceutical, food,animal feed,cosmetic,bio-chemical research etc.

Solubility
It is soluble in methanol. (5mg/ ml-clear colorless solution)

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible to strong oxidizing agents. Stable under recommended storage conditions. Hygroscopic.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7524-50-7
Molecular Formula C10H14ClNO2
Molecular Weight (g/mol) 215.68
MDL Number MFCD00012489
InChI Key SWVMLNPDTIFDDY-UHFFFAOYNA-N
Synonym l-phenylalanine methyl ester hydrochloride, h-phe-ome.hcl, methyl l-phenylalaninate hydrochloride, s-methyl 2-amino-3-phenylpropanoate hydrochloride, unii-47hk4y94ja, h-phe-ome hcl, h-phe-ome hydrochloride, l-phenylalanine, methyl ester, hydrochloride, l-phenylalanine methyl ester hcl
PubChem CID 75736
IUPAC Name methyl (2S)-2-amino-3-phenylpropanoate;hydrochloride
SMILES [H+].[Cl-].COC(=O)C(N)CC1=CC=CC=C1

Specifications

Melting Point 158°C to 162°C
Beilstein 3597948
Solubility Information It is soluble in methanol. (5mg/ ml-clear colorless solution)
Quantity 10 g
Formula Weight 215.68
Optical Rotation +37° (c=2 in ethanol)
Sensitivity Hygroscopic
Percent Purity 99%
Chemical Name or Material L-Phenylalanine methyl ester hydrochloride

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
L-Phenylalanine methyl ester hydrochloride, 99%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.