missing translation for 'onlineSavingsMsg'
Learn More

L(-)-Phenylephrine hydrochloride, 99%, Thermo Scientific Chemicals

99.60 € - 494.46 €

Chemical Identifiers

CAS 61-76-7
Molecular Formula C9H14ClNO2
Molecular Weight (g/mol) 203.67
MDL Number MFCD00012605,MFCD00044749
InChI Key OCYSGIYOVXAGKQ-UHFFFAOYNA-N
Synonym phenylephrine hydrochloride, phenylephrine hcl, l-phenylephrine hydrochloride, metaoxedrine chloride, neosympatol, synethenate, almefrin, consdrin, emagrin, fenilfar
PubChem CID 5284443
ChEBI CHEBI:8094
IUPAC Name 3-[(1R)-1-hydroxy-2-(methylamino)ethyl]phenol;hydrochloride
SMILES [H+].[Cl-].CNCC(O)C1=CC=CC(O)=C1
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10432211
View Documents
Thermo Scientific Acros
207240100
10 g Glass bottle
99.60 €
10g
Please to purchase this item. Need a web account? Register with us today!
10032700
View Documents
Thermo Scientific Acros
207240250
25 g Glass bottle
176.88 €
25g
Please to purchase this item. Need a web account? Register with us today!
10249493
View Documents
Thermo Scientific Acros
207241000
100 g Glass bottle
494.46 €
100g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

61-76-7
203.67
OCYSGIYOVXAGKQ-UHFFFAOYNA-N
5284443
3-[(1R)-1-hydroxy-2-(methylamino)ethyl]phenol;hydrochloride
C9H14ClNO2
MFCD00012605,MFCD00044749
phenylephrine hydrochloride, phenylephrine hcl, l-phenylephrine hydrochloride, metaoxedrine chloride, neosympatol, synethenate, almefrin, consdrin, emagrin, fenilfar
CHEBI:8094
[H+].[Cl-].CNCC(O)C1=CC=CC(O)=C1

Specifications

61-76-7
White
Authentic
C9H14ClNO2
MFCD00012605,MFCD00044749
−47° (20°C c=2,H2O)
phenylephrine hydrochloride, phenylephrine hcl, l-phenylephrine hydrochloride, metaoxedrine chloride, neosympatol, synethenate, almefrin, consdrin, emagrin, fenilfar
[H+].[Cl-].CNCC(O)C1=CC=CC(O)=C1
203.67
CHEBI:8094
99%
L(-)-Phenylephrine hydrochloride
140°C to 145°C
1% max. (1 g, 105°C)
99%
HOC6H4CH(CH2NHCH3)OH·HCl
15, 7398
-47
OCYSGIYOVXAGKQ-UHFFFAOYNA-N
3-[(1R)-1-hydroxy-2-(methylamino)ethyl]phenol;hydrochloride
5284443
203.67
Crystalline Powder
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if swallowed.
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

missing translation for 'einecsNumber' : 200-517-3

Product Identifier
  • L(-)-Phenylephrine hydrochloride

Signal Word
  • Warning

Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.