missing translation for 'onlineSavingsMsg'
Learn More

L-Prolinamide, 98%, Thermo Scientific Chemicals

Product Code. 10227080
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7531-52-4
Molecular Formula C5H11N2O
Molecular Weight (g/mol) 115.16
MDL Number MFCD00005253
InChI Key VLJNHYLEOZPXFW-BYPYZUCNSA-O
Synonym l-prolinamide, prolinamide, h-pro-nh2, s-pyrrolidine-2-carboxamide, s-prolinamide, 2s-pyrrolidine-2-carboxamide, l-prolineamide, l-proline amide, l---prolinamide, s-2-pyrrolidinecarboxamide
PubChem CID 111306
ChEBI CHEBI:21374
IUPAC Name (2S)-pyrrolidine-2-carboxamide
SMILES NC(=O)[C@@H]1CCC[NH2+]1

Specifications

Melting Point 97.0°C to 102.0°C
Color Straw to White
Assay Percent Range 98%
Individual Impurity 97.5% min. (TLC)
Infrared Spectrum Authentic
Packaging Glass Bottle
Specific Rotation − 106.00
Formula Weight 114.15
Percent Purity 98%
Specific Rotation Condition −106° (20°C c=2,EtOH)
Quantity 1 g
Physical Form Powder or Granules
Chemical Name or Material L-Prolinamide
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'