missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals L-(+)-Rhamnose monohydrate, 98+%, Thermo Scientific Chemicals
missing translation for 'orderingAttributeHoverText'
Quantity:
5 g
25 g
100 g
Description
L-(+)-Rhamnose monohydrate is used as a starting material for the production of furanones. It is an important material involved in the reaction of flavors developed during the preparation of various foods like bread, grilled meats, etc. It is also essential to test the permeability of the intestinal tract. Further, it is used as sweeting agent.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
L-(+)-Rhamnose monohydrate is used as a starting material for the production of furanones. It is an important material involved in the reaction of flavors developed during the preparation of various foods like bread, grilled meats, etc. It is also essential to test the permeability of the intestinal tract. Further, it is used as sweeting agent.
Solubility
Soluble in water and methanol. Slightly soluble in ethanol.
Notes
Store in a cool place. Incompatible with strong oxidizing agents.
L-(+)-Rhamnose monohydrate is used as a starting material for the production of furanones. It is an important material involved in the reaction of flavors developed during the preparation of various foods like bread, grilled meats, etc. It is also essential to test the permeability of the intestinal tract. Further, it is used as sweeting agent.
Solubility
Soluble in water and methanol. Slightly soluble in ethanol.
Notes
Store in a cool place. Incompatible with strong oxidizing agents.
Specifications
Melting Point | 89°C to 94°C |
Density | 1.47 |
Quantity | 5 g |
Beilstein | 5988592 |
Merck Index | 14,8172 |
Solubility Information | Soluble in water and methanol. Slightly soluble in ethanol. |
Optical Rotation | +8° (c=10 in Water, 20h) |
IUPAC Name | (2R,3R,4S,5S)-2,3,4,5-tetrahydroxyhexanal;hydrate |
Formula Weight | 182.19 (164.17 Anhydrous) |
Percent Purity | 99% |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'