missing translation for 'onlineSavingsMsg'
Learn More

Masterflex™ L/S™ Variable-Speed Air-Powered Drive with Easy-Load™ II Pump Head

Masterflex™ L/S™ Variable-Speed Air-Powered Drive with Easy-Load™ II Pump Head for use when electrical power is unsafe or unavailable

Brand:  Masterflex™ 77931-10

Product Code. 11731439

  • 2380.00 € / Each
Estimated Shipment: 15-04-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Use where electrical power is unsafe or unavailable
  • Flow range 3.6 to 3400 mL/min - flow rate depends on drive rpm, tubing head, and tubing size
  • Easily vary speed by adjusting air pressure with regulator
  • Rugged 3/4 hp air motor requires little maintenance and generates less heat than an electric motor
  • Careful grounding to protect from static electricity is vital for safe operation. Drive runs on 3 to 25 cfm air input at 20 to 100 psi
  • Air-powered drive is ATEX Zone 2 approved; rated to EEX II 3 G c IIC T6 —drive only, pump head is not ATEX approved
  • IP 44 rated painted steel housing protects from gentle spray
  • Flow range 170 to 1700 mL/min with L/S tubing
  • Includes:
    • Air-Powered Drive (07569-00)
    • Easy-Load™ II Pump Head (77200-62)
    • 10-ft C-Flex™ L/S 24 Tubing
    • Air/Speed Regulator
    • μm Air Filter
    • Automatic Lubricator
    • Pressure Gauge (0 to 30 psi)
    • Muffler
Specifications

Specifications

3.6 to 3400 mL/min.
36.73 cm
L/S™ Variable-Speed Air-Powered Drive (07569-00)
IP44
22.38 cm
For use where electrical power is unsafe or unavailable
12.22 cm
6.9 bar
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.