missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals L-Serine methyleester hydrochloride, 98%

Product Code. 10540584
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5680-80-8
Molecular Formula C4H10ClNO3
Molecular Weight (g/mol) 155.578
MDL Number MFCD00063680
InChI Key NDBQJIBNNUJNHA-DFWYDOINSA-N
Synonym l-serine methyl ester hydrochloride, h-ser-ome.hcl, methyl l-serinate hydrochloride, s-methyl 2-amino-3-hydroxypropanoate hydrochloride, methyl 2s-2-amino-3-hydroxypropanoate hydrochloride, l-serine, methyl ester, hydrochloride, l-methyl serinate hcl, serine methyl ester hydrochloride, l-serine methyl ester hcl, h-ser-ome-hcl
PubChem CID 2723730
IUPAC Name methyl (2S)-2-amino-3-hydroxypropanoate;hydrochloride
SMILES COC(=O)C(CO)N.Cl

Specifications

Melting Point 160.0°C to 165.0°C
Color White
Assay Percent Range 98%
Infrared Spectrum Authentic
Linear Formula HOCH2CH(NH2)CO2CH3·HCl
Beilstein 04, 506
Packaging Glass Bottle
Solubility Information Solubility in water: soluble. Other solubilities: soluble in methanol
Specific Rotation + 4.50
Formula Weight 155.58
Percent Purity 98%
Sulfated Ash 0.1% max.
Water 1% Max. (K.F.)
Specific Rotation Condition + 4.50 (20.00°C c=2,methanol)
Quantity 100 g
Physical Form Crystalline Powder or Crystals
Chemical Name or Material L-Serine methyl ester hydrochloride
Show More Show Less
Product Identifier
  • L-Serine methyl ester hydrochloride

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'