missing translation for 'onlineSavingsMsg'
Learn More

Microspec™ L Shaped Spreaders

Features smooth edges which avoids tearing and digging into agar

Brand:  Microspec™ E4

 View more versions of this product

Product Code. 15752125

  • 605.00 € / Pack of 500
Estimated Shipment: 13-05-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • High Impact Polystyrene construction
  • Full length to avoid contamination
  • Disposable
  • Sterile, aseptically manufactured
  • Ideal for bacterial counts
Specifications

Specifications

Single-use
5.5 in.
Microseal Pouch
180 mm
Spreader
Blue
140 mm
500
High-impact Polystyrene
Aseptically Manufactured
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

One moment while we fetch your results.
Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Microspec™ L Shaped Spreaders > No. per Case: 7500

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.